Skip to Content
Merck
All Photos(1)

Key Documents

RFA452411S-EP

Rotor for Eppendorf® 5424/5424R Centrifuges

FA-45-24-11-Special, aerosol tight, to hold, 24 x 1.5/2.0 mL tubes, lid

Synonym(s):

Lid for Eppendorf® FA-45-48-11 Rotor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103909

feature

lid
to hold 24 x 1.5/2.0 mL tubes
aerosol tight
fixed angle rotor with PTFE-coating

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 022653041 (NA)
Eppendorf® 5424700004 (EU)

parameter

21,130 × g max. RCF

technique(s)

centrifugation: suitable

compatibility

for use with centrifuge models 5424/5424R

Looking for similar products? Visit Product Comparison Guide

Related Categories

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service