Skip to Content
Merck
All Photos(1)

Key Documents

RAB0658

Sigma-Aldrich

Human FETUB / Fetuin-B ELISA Kit

for serum, plasma, cell culture supernatants and urine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type plasma
sample type urine
sample type cell culture supernatant(s)
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 4 ng/mL
standard curve range: 4.39-50 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FETUB(26998)

General description

The antibody pair provided in this kit recognizes human Fetuin B.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABELADEELISA 5X Assay/Sample Diluent Buffer E (Item E2)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mani Mokou et al.
Mediators of inflammation, 2020, 2483435-2483435 (2020-10-17)
Previous studies have suggested that Fetuin-B seems to be a secreted adipokine related to metabolic diseases. However, the results have been inconsistent. Here, our objective is to investigate the changes in circulating Fetuin-B levels in women with polycystic ovary syndrome
Markéta Kovářová et al.
Journal of clinical medicine, 10(3) (2021-01-28)
The hepatokine fetuin A (Fet A) has been associated with diverse pathological states such as insulin resistance, type 2 diabetes, macrovascular disease, and systemic ectopic and vascular calcification. Fet A may also play a role in tumor growth and metastasis.
Li Fang et al.
Journal of assisted reproduction and genetics, 36(6), 1101-1107 (2019-05-11)
To investigate the relationship between serum/follicular fluid fetuin-B levels and fertilization outcomes in conventional IVF cycles. A prospective cohort study of conventional IVF treatments including 78 cycles with low fertilization rates (two pronuclei [2PN] rate < 30%; LF group) and 104 cycles
Jacob M Wozniak et al.
Cell, 182(5), 1311-1327 (2020-09-06)
Staphylococcus aureus bacteremia (SaB) causes significant disease in humans, carrying mortality rates of ∼25%. The ability to rapidly predict SaB patient responses and guide personalized treatment regimens could reduce mortality. Here, we present a resource of SaB prognostic biomarkers. Integrating
Zhibin Li et al.
Journal of diabetes, 10(12), 916-925 (2018-04-20)
This study explored associations of genetic variants in the fetuin B (FETUB) locus with intrahepatic triglyceride (IHTG) content. Four tagging single-nucleotide polymorphisms (SNPs) of the FETUB locus and patatin-like phospholipase domain containing 3 (PNPLA3) rs738409 and transmembrane 6 super family

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service