Skip to Content
Merck
All Photos(2)

Key Documents

RAB0056

Sigma-Aldrich

Mouse MCP-1 / CCL2 ELISA Kit

for cell and tissue lysates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type cell lysate
sample type tissue lysate

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 3 pg/mL
standard curve range: 2.74-2000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Ccl2(20296)

General description

Monocyte chemotactic protein-1 (MCP-1)/C-C motif chemokine ligand 2 (CCL2) is a cytokine, which plays a major role in recruiting macrophages. It is essential for the development of wound healing stages. CCL2 participates in angiogenesis.
The Mouse MCP-1 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse MCP-1 cell lysate and tissue lysate.

Immunogen

Recombinant Mouse MCP1

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABDIL6ELISA 5X Sample Diluent Buffer (Item D2)SDS

  • RABELADEELISA 5X Assay/Sample Diluent Buffer E (Item E2)SDS

  • RABLYSIS12X Cell Lysis Buffer (Item J)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Derek S Whelan et al.
Scientific reports, 10(1), 2642-2642 (2020-02-16)
Mesenchymal stromal cells (MSC) have immunomodulatory effects impacting macrophages, promoting polarisation towards a reparative phenotype. CCL2 is a potent cytokine involved in the recruitment of macrophages. We hypothesised that MSC derived CCL2 may be involved in the MSC therapeutic effect
Fernando J Barreyro et al.
Liver international : official journal of the International Association for the Study of the Liver, 35(3), 953-966 (2014-04-23)
Hepatocyte apoptosis, the hallmark of non-alcoholic steatohepatitis (NASH) contributes to liver injury and fibrosis. Although, both the intrinsic and extrinsic apoptotic pathways are involved in the pathogenesis of NASH, the final common step of apoptosis is executed by a family
Ruozhi Zhao et al.
The Journal of nutritional biochemistry, 25(8), 851-857 (2014-06-14)
Atherosclerotic cardiovascular complications are the leading cause of death in diabetic patients. Monocyte adhesion is an early event for atherogenesis. Previous studies demonstrated that dark-skin berries had cardiovascular protective effects. We hypothesize that Saskatoon berry (SB) powder may reduce monocyte
Zhenyu Yao et al.
Journal of biomedical materials research. Part A, 102(9), 3291-3297 (2013-10-15)
Wear particles generated from total joint replacements can stimulate macrophages to release chemokines, such as monocyte chemoattractant protein 1 (MCP-1), which is the most important chemokine regulating systemic and local cell trafficking and infiltration of monocyte/macrophages in chronic inflammation. One
John E S Park et al.
Thorax, 69(12), 1120-1127 (2014-09-17)
The burden of chronic heart failure (HF) is rising owing to an increased survivorship after myocardial infarction (MI). Pulmonary structural remodelling in patients with HF may protect against oedema while causing dyspnoea, the predominant symptom associated with HF. The cellular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service