Skip to Content
Merck
All Photos(1)

Documents

R4883

Sigma-Aldrich

Resorufin β-D-galactopyranoside

~95%, powder

Synonym(s):

3-Phenoxazone 7-(β-D-galactopyranoside)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H17NO8
CAS Number:
Molecular Weight:
375.33
Beilstein:
4335306
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

product name

Resorufin β-D-galactopyranoside, ~95%

Assay

~95%

form

powder

solubility

DMSO: 20 mg/mL, clear, orange to red

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](Oc2ccc3N=C4C=CC(=O)C=C4Oc3c2)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C18H17NO8/c20-7-14-15(22)16(23)17(24)18(27-14)25-9-2-4-11-13(6-9)26-12-5-8(21)1-3-10(12)19-11/h1-6,14-18,20,22-24H,7H2/t14-,15+,16+,17-,18-/m1/s1

InChI key

QULZFZMEBOATFS-DISONHOPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Resorufin β-D-galactopyranoside is a non-fluorescent compound and is orange-yellow in color. It is hydrolyzed by the enzyme β-galactosidase (β-Gal) to yield fluorescent resorufin.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microchip device for performing enzyme assays
Hadd AG, et al.
Analytical Chemistry, 69(17), 3407-3412 (1997)
M J Eggertson et al.
Biomedical chromatography : BMC, 13(8), 516-519 (1999-12-28)
beta-Galactosidase was incubated for 60 min with the fluorogenic substrate resorufin-beta-D-galactopyranoside, which is converted by the action of the enzyme into resorufin and galactose. A 160 pL aliquot of reaction mixture was analyzed by capillary electrophoresis utilizing laser-induced fluorescence detection.
Glen K Shoemaker et al.
Biochemistry, 42(6), 1707-1710 (2003-02-13)
By use of a capillary electrophoresis-based procedure, it is possible to measure the activity of individual molecules of beta-galactosidase. Molecules from the crystallized enzyme as well as the original enzyme preparation used to grow the crystals both displayed a range
Immobilized enzyme kinetics analyzed by flow-through microfluorimetry: resorufin-beta-D-galactopyranoside as a new fluorogenic substrate for beta-galactosidase
Hofmann J and Sernetz M
Analytica Chimica Acta, 163, 67-72 (1984)
Seung-Yong Jung et al.
Langmuir : the ACS journal of surfaces and colloids, 24(9), 4439-4442 (2008-03-26)
A device with femtoliter-scale chambers and controlled reaction initiation was developed for single-molecule enzymology. Initially separated substrate and enzyme streams were rapidly mixed in a microfluidic device and encapsulated in an array of individual microreactors, allowing for enzyme kinetics to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service