Skip to Content
Merck
All Photos(1)

Key Documents

PZ0380

Sigma-Aldrich

PF-04447943

≥98% (HPLC)

Synonym(s):

1,5-Dihydro-6-[(3S,4S)-4-methyl-1-(2-pyrimidinylmethyl)-3-pyrrolidinyl]-1-(tetrahydro-2H-pyran-4-yl)-4H-pyrazolo[3,4-d]pyrimidin-4-one, 6-[(3S,4S)-4-Methyl-1-(pyrimidin-2-ylmethyl)pyrrolidin-3-yl]-1-(tetrahydro-2H-pyran-4-yl)-1,5-dihydro-4H-pyrazolo[3,4-d]pyrimidin-4-one, PF 4447943

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H25N7O2
CAS Number:
Molecular Weight:
395.46
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

room temp

SMILES string

O=C1C2=C(N(C3CCOCC3)N=C2)N=C([C@H]4[C@H](C)CN(CC5=NC=CC=N5)C4)N1

InChI

1S/C20H25N7O2/c1-13-10-26(12-17-21-5-2-6-22-17)11-16(13)18-24-19-15(20(28)25-18)9-23-27(19)14-3-7-29-8-4-14/h2,5-6,9,13-14,16H,3-4,7-8,10-12H2,1H3,(H,24,25,28)/t13-,16-/m1/s1

InChI key

IWXUVYOOUMLUTQ-CZUORRHYSA-N

Biochem/physiol Actions

PF-04447943 is a potent selective brain penetrant phosphodiesterase PDE9A inhibitor both potency (8 nM) and selectivity (>10 μM versus all the PDEs except PDE1C at >1 μM). It has been investigated for the treatment of cognitive disorders including Alzheimer′s Disease. PF-04447943 increased neurite outgrowth and synapse formation in hippocampal neurons in culture, exhibited synaptic stabilization in an amyloid precursor protein (APP) transgenic mouse model, and improved memory and enhanced attention in several animal models.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patrick R Verhoest et al.
Journal of medicinal chemistry, 55(21), 9045-9054 (2012-07-12)
6-[(3S,4S)-4-Methyl-1-(pyrimidin-2-ylmethyl)pyrrolidin-3-yl]-1-(tetrahydro-2H-pyran-4-yl)-1,5-dihydro-4H-pyrazolo[3,4-d]pyrimidin-4-one (PF-04447943) is a novel PDE9A inhibitor identified using parallel synthetic chemistry and structure-based drug design (SBDD) and has advanced into clinical trials. Selectivity for PDE9A over other PDE family members was achieved by targeting key residue differences between the
Dávid Nagy et al.
Experimental neurology, 263, 122-131 (2014-10-16)
Several neurophysiological abnormalities have been described in Huntington's disease, including auditory gating deficit, which are considered to reflect impaired brain information-processing. Since transgenic animal models of Huntington's disease capture basic neuropathology of the disorder, auditory gating was studied in BACHD
Ryan T Cameron et al.
FEBS open bio, 7(1), 64-73 (2017-01-18)
Phosphodiesterase (PDE) inhibitors are currently under evaluation as agents that may facilitate the improvement of cognitive impairment associated with Alzheimer's disease. Our aim was to determine whether inhibitors of PDEs 4, 5 and 9 could alleviate the cytotoxic effects of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service