Skip to Content
Merck
All Photos(1)

Documents

P8727

Sigma-Aldrich

Palmityl trifluoromethyl ketone

phosholipase A2 inhibitor

Synonym(s):

PACOCF3, Pentadecyl trifluoromethyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H31F3O
CAS Number:
Molecular Weight:
308.42
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Assay

≥98% (TLC)

form

liquid

concentration

10 mg/mL in ethanol

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)C(F)(F)F

InChI

1S/C17H31F3O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(21)17(18,19)20/h2-15H2,1H3

InChI key

MAHYXYTYTLCTQD-UHFFFAOYSA-N

Biochem/physiol Actions

Reversible inhibitor of calcium-dependent phospholipase A2 (IC50 = 3.8 μM). Unlike other PLA2 inhibitors studied, PACOCF3 at 50 μM induced an increase in intracellular [Ca2+], and at 10 μM, enhanced calcium-ion influx induced by ATP and bradykinin.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dual action of palmitoyl trifluoromethyl ketone (PACOCF3) on Ca2+ signaling: activation of extracellular Ca2+ influx and alteration of ATP- and bradykinin-induced Ca2+ responses in Madin Darby canine kidney cells
Jan, C.R., et al.
Toxicology, 74, 447-451 (2000)
Oscar Yanes et al.
Nature chemical biology, 6(6), 411-417 (2010-05-04)
Metabolites offer an important unexplored complementary approach to understanding the pluripotency of stem cells. Using MS-based metabolomics, we show that embryonic stem cells are characterized by abundant metabolites with highly unsaturated structures whose levels decrease upon differentiation. By monitoring the
Susann Liebscher et al.
PLoS pathogens, 14(4), e1007029-e1007029 (2018-05-01)
Positive-sense RNA virus intracellular replication is intimately associated with membrane platforms that are derived from host organelles and comprised of distinct lipid composition. For flaviviruses, such as West Nile virus strain Kunjin virus (WNVKUN) we have observed that these membrane
Y C Lio et al.
Biochimica et biophysica acta, 1302(1), 55-60 (1996-07-12)
Methyl arachidonyl fluorophosphonate (MAFP) has been recently reported to be a selective, active-site directed, irreversible inhibitor of the Group IV 85 kDa cytosolic phospholipase A2 (cPLA2). We have now shown that this compound also potently inhibits the Ca(2+)-independent cytosolic phospholipase
E J Ackermann et al.
The Journal of biological chemistry, 270(1), 445-450 (1995-01-06)
A novel Ca(2+)-independent phospholipase A2 (PLA2) has recently been purified from the murine macrophage-like cell line P388D1 (Ackermann, E. J., Kempner, E. S., and Dennis, E. A. (1994) J. Biol. Chem. 269, 9227-9233). This enzyme is now shown to be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service