P7780
Propionylpromazine hydrochloride
Synonym(s):
Propiopromazine hydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C20H24N2OS · HCl
CAS Number:
Molecular Weight:
376.94
Beilstein:
38502
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Recommended Products
form
powder
Quality Level
storage temp.
2-8°C
SMILES string
Cl.CCC(=O)c1ccc2Sc3ccccc3N(CCCN(C)C)c2c1
InChI
1S/C20H24N2OS.ClH/c1-4-18(23)15-10-11-20-17(14-15)22(13-7-12-21(2)3)16-8-5-6-9-19(16)24-20;/h5-6,8-11,14H,4,7,12-13H2,1-3H3;1H
InChI key
ZFWVWZODBGTOIL-UHFFFAOYSA-N
Gene Information
human ... DRD2(1813)
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Phenothiazine antipsychotic; D2 dopamine receptor antagonist.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Y Govaert et al.
The Analyst, 123(12), 2507-2512 (1999-08-06)
A rapid and sensitive multi-residue method was developed to attempt to confirm the presence of the beta-blocker carazolol and the tranquillizers acepromazine, azaperone, chlorpromazine, propionylpromazine and xylazine in pig muscle tissues. The procedure involves determination by liquid chromatography coupled with
Residues of some veterinary drugs in animals and foods. Monographs prepared by the thirty-eighth meeting of the Joint FAO/WHO Expert Committee on Food Additives.
FAO food and nutrition paper, 41(4), 1-136 (1991-01-01)
[The opinion of O. Habler].
O Habler
Der Anaesthesist, 50(8), 613-614 (2001-09-15)
[Acute normovolemic hemodilution (ANH). Der Anaesthesist (2000) 49: 939-948].
M Alef
Der Anaesthesist, 50(8), 612-613 (2001-09-15)
[Effects and side effects of some combinations of often used tranquillizers for sedation in horses].
W Rohr et al.
DTW. Deutsche tierarztliche Wochenschrift, 89(7), 262-267 (1982-07-06)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service