Skip to Content
Merck
All Photos(1)

Key Documents

P2519

Sigma-Aldrich

L-Pipecolic acid

99% (titration), suitable for GC/MS

Synonym(s):

(S)-(−)-2-Piperidinecarboxylic acid, L-Homoproline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NO2
CAS Number:
Molecular Weight:
129.16
Beilstein:
81093
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.26

product name

L-Pipecolic acid, 99% (titration)

Assay

99% (titration)

form

powder

technique(s)

GC/MS: suitable

color

white

mp

272 °C (lit.)

SMILES string

OC(=O)[C@@H]1CCCCN1

InChI

1S/C6H11NO2/c8-6(9)5-3-1-2-4-7-5/h5,7H,1-4H2,(H,8,9)/t5-/m0/s1

InChI key

HXEACLLIILLPRG-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-pipecolic acid (piperidine-2-carboxylic acid) is a nitrogen-containing heterocyclic compound. It is an imino acid. Its molecular weight is 129.2.

Application

L-Pipecolic acid has been used as a standard for the quantification by gas chromatography-mass spectrometry (GC-MS).

Biochem/physiol Actions

L-pipecolic acid is implicated in Zellweger syndrome. Pipecolic acid is also a plant defence metabolite. It is a signaling compound essential for systemic acquired resistance (SAR).
L-Pipecolic acid is a lysine metabolite; defect in its catabolism is involved in hyperpipecolic acidemia, cerebro-hepato-renal syndrome, neonatal onset adrenoleukodystrophy, and infantile Refsum disease.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pipecolic acid
Laboratory Guide to the Methods in Biochemical Genetics (2008)
Transcriptional analysis of defense mechanisms in upland tetraploid switchgrass to greenbugs
Donze-Reiner T, et al.
BMC plant biology, 17(1), 46-46 (2017)
S J Mihalik et al.
The Journal of biological chemistry, 264(5), 2509-2517 (1989-02-15)
L-Pipecolic acid oxidation was studied in the rabbit and cynomolgus monkey. Tissue homogenates from both species incubated with L-[2,3,4,5,6-3H]pipecolic acid produced a single radioactive product identified as alpha-aminoadipic acid. In the rabbit, L-pipecolic acid oxidation was greatest in kidney cortex
M R Baumgartner et al.
Annals of neurology, 47(1), 109-113 (2000-01-13)
We describe an 18-year-old patient with psychomotor retardation and abnormally short metatarsi and metacarpals but no other signs of classic Refsum disease. Molecular analysis of the phytanoyl-coenzyme A hydroxylase gene revealed a homozygous deletion causing a frameshift. Surprisingly, L-pipecolic acid
S J Mihalik et al.
Pediatric research, 25(5), 548-552 (1989-05-01)
L-Pipecolic acid, a cyclic imino acid produced during the degradation of lysine, accumulates in body fluids of infants with the generalized peroxisomal disorders, including Zellweger syndrome, neonatal adrenoleukodystrophy, and infantile Refsum disease. Peroxisome-enriched fractions from normal human liver oxidized L-[3H]pipecolic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service