P1110
Dounce tissue grinder pestle
Small clearance, working volume 2 mL
Synonym(s):
885302-0002
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
eCl@ss:
32080115
NACRES:
NB.22
Recommended Products
material
glass
working volume
2 mL
Looking for similar products? Visit Product Comparison Guide
General description
Designed primarily for cellular work where the nucleus remains intact after homogenization.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Lukas Minus Orre et al.
Molecular cell, 73(1), 166-182 (2019-01-05)
Subcellular localization is a main determinant of protein function; however, a global view of cellular proteome organization remains relatively unexplored. We have developed a robust mass spectrometry-based analysis pipeline to generate a proteome-wide view of subcellular localization for proteins mapping
Xiaotong Yang et al.
Stem cell research & therapy, 11(1), 182-182 (2020-05-21)
ARDS and ALI are life-threatening diseases with extremely high mortality in patients. Different sources of MSCs could mitigate the symptoms of ALI from diverse mechanisms. Liraglutide is an activator of glucagon-like peptide-1 receptor (GLP-1R) that activates anti-apoptotic pathways and exerts
Feifei Ren et al.
Immunology and cell biology, 97(5), 457-469 (2018-12-24)
Mutations in the isocitrate dehydrogenase (IDH) 1 gene, especially the R132H mutation, have been reported to be associated with a better prognosis in glioma patients. However, the underlying molecular mechanisms are not yet well understood. Many factors may contribute to
Jinyu Zhang et al.
Oncology letters, 19(1), 350-358 (2020-01-04)
Cisplatin (DDP) resistance is closely associated with the failure of chemotherapy to manage various different types of human cancer. The GTPase protein Ras-related protein Rap-2a (RAP2A) regulates cancer cell proliferation, migration and invasion; however, little is currently known regarding its
Hao Zhang et al.
Journal of animal science, 98(10) (2020-10-08)
This investigation evaluated the potential of natural antioxidants, pterostilbene (PT) and its parent compound resveratrol (RSV), to alleviate hepatic damage, redox imbalance, mitochondrial dysfunction, and endoplasmic reticulum (ER) stress in early-weaned piglets. A total of 144 suckling piglets were randomly
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service