Skip to Content
Merck
All Photos(1)

Key Documents

N9910

Sigma-Aldrich

β-Nicotinamide adenine dinucleotide 2′-phosphate reduced tetrasodium salt

vial of 0.30-0.36 mg

Synonym(s):

β-NADPH, 2′-NADPH, Coenzyme II reduced tetrasodium salt, Dihydronicotinamide adenine dinucleotide phosphate tetrasodium salt, NADPH Na4, TPNH2 Na4, Triphosphopyridine nucleotide reduced tetrasodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26N7Na4O17P3
Molecular Weight:
833.35
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51
Pricing and availability is not currently available.

form

solid

Quality Level

packaging

vial of 0.30-0.36 mg

SMILES string

[Na+].[Na+].[Na+].[Na+].[P](=O)([O-])([O-])O[C@H]1[C@@H](O[C@@H]([C@H]1O)CO[P](=O)([O-])O[P](=O)([O-])OC[C@@H]4O[C@@H]([C@H]([C@H]4O)O)N5C=C(CC=C5)C(=O)N)[n]2c3ncnc(c3nc2)N

InChI

1S/C21H30N7O17P3.4Na/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(44-46(33,34)35)14(30)11(43-21)6-41-48(38,39)45-47(36,37)40-5-10-13(29)15(31)20(42-10)27-3-1-2-9(4-27)18(23)32;;;;/h1,3-4,7-8,10-11,13-16,20-21,29-31H,2,5-6H2,(H2,23,32)(H,36,37)(H,38,39)(H2,22,24,25)(H2,33,34,35);;;;/q;4*+1/p-4/t10-,11+,13-,14+,15-,16+,20-,21+;;;;/m0..../s1

InChI key

WYWWVJHQDVCHKF-MPUNMZHWSA-J

Looking for similar products? Visit Product Comparison Guide

Application

β-Nicotinamide adenine dinucleotide 2′-phosphate (NADP+ and β-Nicotinamide adenine dinucleotide 2′-phosphate, reduced (NADPH) comprise a coenzyme redox pair (NADP+:NADPH) involved in a wide range of enzyme catalyzed oxidation reduction reactions. The NADP+/NADPH redox pair facilitates electron transfer in anabolic reactions such as lipid and cholesterol biosynthesis and fatty acyl chain elongation. The NADP+/NADPH redox pair is used in a variety of antioxidation mechanism where it protects agains reactive oxidation species accumulation. NADPH is generated in vivo by the pentose phosphate pathway (PPP).

Biochem/physiol Actions

β-Nicotinamide adenine dinucleotide 2′-phosphate (NADP+) and β-Nicotinamide adenine dinucleotide 2′-phosphate, reduced (NADPH) comprise a coenzyme redox pair (NADP+:NADPH) involved in a wide range of enzyme catalyzed oxidation reduction reactions. The NADP+/NADPH redox pair facilitates electron transfer in anabolic reactions such as lipid and cholesterol biosynthesis and fatty acyl chain elongation. The NADP+/NADPH redox pair is used in a variety of antioxidation mechanism where it protects agains reactive oxidation species accumulation. NADPH is generated in vivo by the pentose phosphate pathway (PPP).
Electron donor; cofactor for many redox enzymes including nitric oxide synthetase

Preparation Note

Chemically reduced

Other Notes

Packaged based on NADPH content as determined by UV-Vis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zoltán Serfőző et al.
Cellular signalling, 30, 67-81 (2016-11-26)
Olfaction, a chemosensory modality, plays a pivotal role in the orientation and behavior of invertebrates. The central olfactory processing unit in terrestrial stylomatophoran snails is the procerebrum, which contains NO synthesizing interneurons, whose oscillatory currents are believed to be the
Zoltán Serfőző et al.
Cellular signalling, 30, 67-81 (2016-11-26)
Olfaction, a chemosensory modality, plays a pivotal role in the orientation and behavior of invertebrates. The central olfactory processing unit in terrestrial stylomatophoran snails is the procerebrum, which contains NO synthesizing interneurons, whose oscillatory currents are believed to be the
Laura Canevari et al.
Neurochemical research, 32(4-5), 739-750 (2006-12-28)
Since the discovery of the significance of the cholesterol-carrying apolipoprotein E and cholesterolaemia as major risk factors for Alzheimer's Disease (AD) there has been a mounting interest in the role of this lipid as a possible pathogenic agent. In this
Estrogen signaling and neuroprotection in cerebral ischemia.
Brann D, Raz L, Wang R, et al.
Journal of Neuroendocrinology, 24(1), 34-47 (2011)
Paul A Fraser
Free radical biology & medicine, 51(5), 967-977 (2011-06-30)
The brain endothelium constitutes a barrier to the passive movement of substances from the blood into the cerebral microenvironment, and disruption of this barrier after a stroke or trauma has potentially fatal consequences. Reactive oxygen species (ROS), which are formed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service