Skip to Content
Merck
All Photos(1)

Key Documents

N7255

Sigma-Aldrich

1-Naphthyl phosphate disodium salt

phosphatase substrate, fluorogenic, powder

Synonym(s):

α-Naphthyl phosphate disodium salt, Disodium 1-naphthyl phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7Na2O4P
CAS Number:
Molecular Weight:
268.11
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Product Name

1-Naphthyl phosphate disodium salt, phosphatase substrate

Quality Level

form

powder

composition

phosphorous, 11.2-12.4% ICP (anhydrous)
sodium, 16.0-18.4% ICP (anhydrous)

solubility

water: 50 mg/mL, clear, colorless to very faintly yellow

storage temp.

−20°C

SMILES string

[Na+].[Na+].[O-]P([O-])(=O)Oc1cccc2ccccc12

InChI

1S/C10H9O4P.2Na/c11-15(12,13)14-10-7-3-5-8-4-1-2-6-9(8)10;;/h1-7H,(H2,11,12,13);;/q;2*+1/p-2

InChI key

QYURIFWAOPAPAJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

1-Naphthyl phosphate disodium salt has been used:
  • as a component in the mixture for soaking the chromatography paper for determining acid phosphatase activity
  • in the staining of the alkaline phosphatase isoforms in 7.5% non-denaturing polyacrylamide gel
  • as a component in the mixture for soaking the chromatography paper for preparing acid phosphomonoesterase-reactive imprinting sheets
  • as a component in the staining solution for staining the roots for determining phosphomonoesterase activity

Substrates

Substrate for determination of phosphatase.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A method for linking in situ activities of hydrolytic enzymes to associated organisms in forest soils
Dong S, et al.
Soil Biology and Biochemistry, 39(9), 2414-2419 (2007)
The net charge of the first 18 residues of the mature sequence affects protein translocation across the cytoplasmic membrane of gram-negative bacteria
Kajava AV, et al.
Journal of Bacteriology, 182(8), 2163-2169 (2000)
Improving phosphorus acquisition of white clover (Trifolium repens L.) by transgenic expression of plant-derived phytase and acid phosphatase genes
Ma XF, et al.
Plant Science, 176(4), 479-488 (2009)
Processing of Escherichia coli Alkaline Phosphatase SEQUENCE REQUIREMENTS AND POSSIBLE CONFORMATIONS OF THE- 6 TO- 4 REGION OF THE SIGNAL PEPTIDE
Kajava AV, et al.
The Journal of Biological Chemistry, 277(52), 50396-50402 (2002)
Nambirajan Rangarajan et al.
The Journal of biological chemistry, 295(41), 14189-14202 (2020-08-14)
Autophagy is a conserved process that recycles cellular contents to promote survival. Although nitrogen limitation is the canonical inducer of autophagy, recent studies have revealed several other nutrients important to this process. In this study, we used a quantitative, high-throughput

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service