Skip to Content
Merck
All Photos(5)

Key Documents

N5879

Sigma-Aldrich

O-(4-Nitrophenylphosphoryl)choline

Synonym(s):

p-Nitrophenylphosphorylcholine, NPPC, Phosphocholine 4-nitrophenyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H17N2O6P
CAS Number:
Molecular Weight:
304.24
Beilstein:
1895422
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

Quality Level

storage temp.

−20°C

SMILES string

C[N+](C)(C)CCOP([O-])(=O)Oc1ccc(cc1)[N+]([O-])=O

InChI

1S/C11H17N2O6P/c1-13(2,3)8-9-18-20(16,17)19-11-6-4-10(5-7-11)12(14)15/h4-7H,8-9H2,1-3H3

InChI key

NAIXASFEPQPICN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reported to be useful in assay of Phospholipase C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dolores Pérez et al.
Food microbiology, 97, 103763-103763 (2021-03-04)
A collection of 33 Saccharomyces yeasts were used for wine fermentation with a sole nitrogen source: ammonium and four individual aroma-inducing amino acids. The fermentation performance and chemical wine composition were evaluated. The most valuable nitrogen sources were valine as
Michael V Airola et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(28), E5549-E5558 (2017-06-28)
Neutral sphingomyelinase 2 (nSMase2, product of the
Wan-Chun Li et al.
Scientific reports, 8(1), 8212-8212 (2018-05-31)
Ductal epithelium is primarily detected in porcine neonatal pancreatic cell clusters (NPCCs) bearing grafts, suggesting that transplants might exhibit progenitor-like phenotypes. Here we found that soon after NPCC isolation, PDX1+/insulin- and SOX9+ pancreatic progenitor-like cells dramatically increased while dual-hormonal progenitor-like
New York City Panel on Climate Change 2019 Report Chapter 5: Mapping Climate Risk.
Lesley Patrick et al.
Annals of the New York Academy of Sciences, 1439(1), 115-125 (2019-03-16)
María Belén Mazzucco et al.
Antonie van Leeuwenhoek, 112(7), 965-973 (2019-01-24)
The juicing industry generates large amounts of waste that mostly lack commercial value and, in the absence of waste treatment policies, produces environmental pollution. Also, microbiological spoilage is a major concern in the wine industry and control tools are limited.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service