Skip to Content
Merck
All Photos(1)

Key Documents

MAK136

Sigma-Aldrich

Monoamine Oxidase Activity Assay Kit

sufficient for 100 fluorometric tests

Synonym(s):

MAO Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 fluorometric tests

detection method

fluorometric

relevant disease(s)

neurological disorders

storage temp.

−20°C

Gene Information

General description

Monoamine oxidases (MAO, EC 1.4.3.4) are a family of mitochondrial enzymes that catalyze the oxidative deamination of monoamines. In mammals, MAO consists of two isoenzymes, MAO-A and MAO-B. MAO dysfunction is thought to be responsible for a number of neurological disorders. Unusually high or low levels of MAO in the body have been associated with depression, schizophrenia, substance abuse, attention deficit disorder, migraines, and irregular sexual maturation. MAO inhibitors are one of the major classes of drug prescribed for the treatment of depression.

Application

Monoamine Oxidase Activity Assay Kit has been used to estimate MAO-A (monoamine oxidase A), MAO-B (monoamine oxidase B) and ALDH (aldehyde dehydrogenase) activities.

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

Suitable for the determination of MAO activity in biological samples and for the screening of MAO inhibitors. Can determine either MAO-A, MAO-B or both MAO-A and MAO-B activity.

Principle

The Monoamine Oxidase Assay Kit provides a convenient fluorimetric means to measure MAO enzyme activity in biological samples. In the assay, MAO reacts with p-tyramine, a substrate for both MAO-A and MAO-B, resulting in the formation of H2O2, which is determined by a fluorimetric method (λex = 530/λem = 585 nm). The assay is simple, sensitive, stable, and high-throughput adaptable. Unit definition: one unit of MAO catalyzes the formation of 1 μmole of H2O2 per minute under the assay conditions.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lovedeep Singh et al.
Metabolic brain disease, 36(1), 111-121 (2020-09-02)
Pain and depression are often co-existing pathological states that promote mutual severity resulting in limited efficacy of current treatment strategies. Thus, there is a need to develop an efficacious alternate treatment regimen for pain-depression dyad. Skimmetin and osthole are molecules
Proteome Profiling of Primary Pancreatic Ductal Adenocarcinomas Undergoing Additive Chemoradiation Link ALDH1A1 to Early Local Recurrence and Chemoradiation Resistance.
Oria V O, et al.
Translational Oncology, 11(6), 1307-1322 (2018)
Umut Salgin-Goksen et al.
Journal of medicinal chemistry, 64(4), 1989-2009 (2021-02-04)
Thirty compounds having 1-[2-(5-substituted-2-benzoxazolinone-3-yl) acetyl]-3,5-disubstitutedphenyl-2-pyrazoline structure and nine compounds having N'-(1,3-disubstitutedphenylallylidene)-2-(5-substituted-2-benzoxazolinone-3-yl)acetohydrazide skeleton were synthesized and evaluated as monoamine oxidase (MAO) inhibitors. All of the compounds exhibited selective MAO-A inhibitor activity in the nanomolar or low micromolar range. The results of
Juho-Matti Renko et al.
Molecular neurobiology, 55(8), 6755-6768 (2018-01-20)
Neurotrophic factors (NTFs) hold potential as disease-modifying therapies for neurodegenerative disorders like Parkinson's disease. Glial cell line-derived neurotrophic factor (GDNF), cerebral dopamine neurotrophic factor (CDNF), and mesencephalic astrocyte-derived neurotrophic factor (MANF) have shown neuroprotective and restorative effects on nigral dopaminergic
Mesencephalic Astrocyte-Derived Neurotrophic Factor (MANF) Elevates Stimulus-Evoked Release of Dopamine in Freely-Moving Rats.
Renko J M, et al.
Molecular Neurobiology, 1-14 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service