Skip to Content
Merck
All Photos(2)

Key Documents

M7693

Sigma-Aldrich

Anti-MTA1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Metastasis-associated Gene, Anti-NuRD-70

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 75-80 kDa (doublet)

species reactivity

human

technique(s)

immunoprecipitation (IP): 5-10 μg/mL using extracts of 293T cells
microarray: suitable
western blot: 0.5-1 μg/mL using nuclear extracts of MCF7 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MTA1(9112)

General description

MTA1 gene is mapped to human chromosome 14q32.33.
Metastasis associated 1 (MTA1) is a 80kDa protein which possesses four domains and several interaction motifs. It is made up of 715 amino acid residues.

Specificity

Anti-MTA1 recognizes specifically MTA1.

Immunogen

synthetic peptide corresponding to amino acids 626-641 of human MTA1, conjugated to KLH via an N-terminal added cysteine residue. The immunizing sequence is not present in other members of the family, MTA2 and MTA3.

Application

Anti-MTA1 antibody produced in rabbit has been used in:
  • western blotting
  • immunohistochemistry
  • immunoprecipitation.

Biochem/physiol Actions

MTA1 interacts with cyclin-dependent kinase-activating kinase (CAK) is a component of the transcription factor II H (TFIIH) regulatory complex, suggesting its role as a signal transducer to mediate crosstalk between corepressor complexes and the general transcription machinery.
Metastasis associated 1 (MTA1) has been shown to be overexpressed in many types of cancers and has an important role in cancer malignancy. It takes part in the DNA repair process. As part of the nuclear remodeling and deacetylation complex (NuRD complex), it associates with deacetylated p53. MTA1 participates as a co-repressor and co-activator for many kinds of genes.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. Storage in frost-free freezers is also not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilu-tions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher J Millard et al.
Cancer metastasis reviews, 33(4), 857-867 (2014-10-30)
Gene expression is controlled through the recruitment of large coregulator complexes to specific gene loci to regulate chromatin structure by modifying epigenetic marks on DNA and histones. Metastasis-associated protein 1 (MTA1) is an essential component of the nucleosome remodelling and
De novo unbalanced translocation resulting in monosomy for distal 5p (5p14. 1? pter) and 14q (14q32. 31? qter) associated with fetal nuchal edema, microcephaly, intrauterine growth restriction, and single umbilical artery: prenatal diagnosis and molecular cytogenetic characterization
Chen CP, et al.
Taiwanese Journal of Obstetrics & Gynecology, 52(3), 401-406 (2013)
Involvement of metastasis tumor antigen 1 in hepatic regeneration and proliferation
Li W, et al.
Cellular Physiology and Biochemistry, 22(1-4), 315-326 (2008)
Ekjot Kaur et al.
Cancer metastasis reviews, 33(4), 1017-1024 (2014-11-07)
Metastasis-associated gene or metastasis tumor antigen 1 (MTA1) is a new member of cancer progression-related gene family. It was first identified in rat mammary adenocarcinoma and later recognized as an important constituent of nucleosomal remodeling complex (NuRD), displaying dual regulatory
MTA1 interacts with MAT1, a cyclin-dependent kinase-activating kinase complex ring finger factor, and regulates estrogen receptor transactivation functions
Talukder AH, et al.
The Journal of Biological Chemistry, 278(13), 11676-11685 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service