Skip to Content
Merck
All Photos(4)

Documents

L8021

Sigma-Aldrich

D-Lysine

≥98% (HPLC)

Synonym(s):

(R)-2,6-Diaminocaproic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2N(CH2)4CH(NH2)CO2H
CAS Number:
Molecular Weight:
146.19
Beilstein:
1722530
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

D-Lysine, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

color

white to off-white

mp

218 °C (dec.) (lit.)

solubility

H2O: soluble

storage temp.

−20°C

SMILES string

NCCCC[C@@H](N)C(O)=O

InChI

1S/C6H14N2O2/c7-4-2-1-3-5(8)6(9)10/h5H,1-4,7-8H2,(H,9,10)/t5-/m1/s1

InChI key

KDXKERNSBIXSRK-RXMQYKEDSA-N

Looking for similar products? Visit Product Comparison Guide

Application

D-Lysine is used as a component of a wide array of polymers (poly-D-lysine), surfactants and biofilms to confer a positive (cationic) charge.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isabel López-García et al.
British journal of pharmacology, 175(2), 284-300 (2016-10-11)
We hypothesized that an in vitro, stretch-based model of neural injury may be useful to identify compounds that decrease the cellular damage in neurotrauma. We screened three neural cell lines (B35, RN33B and SH-SY5Y) subjected to two differentiation methods and
Craig A Erickson et al.
Journal of psychiatric research, 84, 153-160 (2016-10-16)
Dysregulation of extracellular signal-related kinase (ERK) activity has been potentially implicated in the pathophysiology of autistic disorder (autism). ERK is part of a central intracellular signaling cascade responsible for a myriad of cellular functions. ERK is expressed in peripheral blood
Sara Fornera et al.
Macromolecular bioscience, 11(8), 1052-1067 (2011-05-14)
Horseradish peroxidase (HRP) is immobilized in three easy steps on SiO(2) surfaces with the help of a polycationic second generation dendronized polymer (denpol) and the biotin-avidin system. This stepwise immobilization process is monitored and quantitatively analyzed with the transmission interferometric
S Lule et al.
Journal of autoimmunity, 84, 87-96 (2017-08-29)
Autoimmune and dysimmune inflammatory mechanisms on a genetically susceptible background are implicated in the etiology of Behçet's Disease (BD). Heat-shock protein-65 (HSP-65) derived from Streptococcus sanguinis was proposed as a triggering factor based on its homology with human HSP-60. However
Daniel Martin et al.
Nature communications, 9(1), 2372-2372 (2018-07-10)
Dysregulation of the Hippo signaling pathway and the consequent YAP1 activation is a frequent event in human malignancies, yet the underlying molecular mechanisms are still poorly understood. A pancancer analysis of core Hippo kinases and their candidate regulating molecules revealed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service