Skip to Content
Merck
All Photos(2)

Documents

L7025

Sigma-Aldrich

LB Broth with agar (Lennox)

Tablet microbial growth medium

Synonym(s):

LB Agar, LB agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.85

grade

for molecular biology

sterility

non-sterile

form

tablet

technique(s)

microbiological culture: suitable

application(s)

food and beverages
microbiology

storage temp.

room temp

suitability

nonselective for Escherichia coli
nonselective for coliforms

General description

Lennox LB is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides, amino acids, water-soluble vitamins, and carbohydrates in a low-salt formulation. The addition of agar provides a solid medium for microbial growth.

Application

Suitable for non-selective cultivation of E. coli strains for cloning, DNA plasmid production and production of recombinant proteins. Also suitable for selective cultivation when appropriate antibiotics are added, including those that require low-salt conditions, such as Zeocin®.

Features and Benefits

Lennox LB tablet with agar provides:
  • A budget-friendly alternative to pre-poured plates
  • Convenient small package to eliminate weighing
  • Fast preparation – no stirring required

Components

13.72g/L Agar
9.14g/L Tryptone
4.57 g/L Yeast Extract
4.57 g/L NaCl
1.6g/L inert binding agents

Reconstitution

Add each 1.68 g tablet to 48.3 mL water (no stirring required). Autoclave for 20 minutes at 121 °C to sterilize. Allow to cool slightly before making additions, such as antibiotics (if desired). Pour into petri dishes and allow to solidify.

Legal Information

Zeocin is a registered trademark of Cayla Sarl

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shun Gen Huang et al.
BMC immunology, 14, 27-27 (2013-06-22)
Surgical intervention-related trauma contributes largely to the development of postoperative immunosuppression, with reduced resistance to secondary bacterial infection. This study compared the impact of laparotomy versus laparoscopy on macrophage-associated bactericidal ability and examined whether laparotomy renders the host more susceptible
Wenjun Kong et al.
Nature protocols, 15(3), 750-772 (2020-02-14)
Single-cell technologies are offering unparalleled insight into complex biology, revealing the behavior of rare cell populations that are masked in bulk population analyses. One current limitation of single-cell approaches is that lineage relationships are typically lost as a result of
Atlas, R. M., L.C. Parks
Handbook of Microbiological Media (1993)

Protocols

General protocols for growth of competent cells in microbial medium.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service