Skip to Content
Merck
All Photos(3)

Documents

L6001

Sigma-Aldrich

DL-Lysine monohydrochloride

≥98% (HPLC)

Synonym(s):

DL-2,6-Diaminohexanoic acid monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2(CH2)4CH(NH2)COOH · HCl
CAS Number:
Molecular Weight:
182.65
Beilstein:
4711993
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

DL-Lysine monohydrochloride, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

color

white to off-white

mp

267 °C (dec.) (lit.)

solubility

H2O: soluble

application(s)

peptide synthesis

SMILES string

Cl.NCCCCC(N)C(O)=O

InChI

1S/C6H14N2O2.ClH/c7-4-2-1-3-5(8)6(9)10;/h5H,1-4,7-8H2,(H,9,10);1H

InChI key

BVHLGVCQOALMSV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

DL-Lysine monohydrochloride is a racemic mixture of the D- and L- enantiomers of lysine. DL-Lysine may be used to construct polylysine polymers useful to produce lysine-based polypeptide gels and films.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mamoru K Kokufuta et al.
Colloids and surfaces. B, Biointerfaces, 87(2), 299-309 (2011-06-21)
We synthesized the glutaraldehyde cross-linked hydrogels using four kinds of poly(lysine)s (PLs) and measured the equilibrium swelling ratio (Q) as a function of pH. Also measured was the temperature change of Q at a fixed pH (11.6) in the absence
K Itoh et al.
Langmuir : the ACS journal of surfaces and colloids, 24(23), 13426-13433 (2008-11-01)
The buildup mechanism of polypeptide multilayers prepared by the layer-by-layer deposition of a polyanion (poly(L-glutamic acid) (PGA)) and polycations (poly(L-lysine) (PLL), poly(D-lysine) (PDL), and copoly(DL-lysine)(PDLL)) was reinvestigated by using in situ ATR-IR spectroscopy. A difference spectral technique applied to analyze
Preparation of hierarchical mesocrystalline DL-lysine·HCl-poly(acrylic acid) hybrid thin films.
Yuan Jiang et al.
Advanced materials (Deerfield Beach, Fla.), 23(31), 3548-3552 (2011-07-05)
Pasquale De Santis et al.
Journal of nanoscience and nanotechnology, 7(7), 2230-2238 (2007-08-01)
Organic trans-annular assemblies constitute an expanding class of structures with promising applications for the design of nanotechnological devices. Among the strategies developed for the engineering of organic nanotubes, those characterized by regular alternating enantiomeric amino acid sequences have been proven
Moyu Taniguchi et al.
Journal of bioscience and bioengineering, 124(4), 414-418 (2017-06-02)
Although naturally abundant amino acids are represented mainly by l-enantiomers, fermented foods are known to contain various d-amino acids. Enantiospecific profiles of food products can vary due to fermentation by bacteria, and such alterations may contribute to changes in food

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service