Skip to Content
Merck
All Photos(2)

Key Documents

K3627

Sigma-Aldrich

Kallikrein from porcine pancreas

≥40 units/mg protein

Synonym(s):

Kininogenase, Kininogenin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

form

solid

Quality Level

specific activity

≥40 units/mg protein

storage temp.

2-8°C

General description

Kallikrein exists as an inactive prokallikrein in the porcine pancreas.[1] The porcine kallikrein gene region is localized on chromosome 6q12-q21.[2]

Application

Kallikrein from porcine pancreas has been used:
  • as a matrix metalloproteinase-9 (MMP-9) zymogen activator[3]
  • as a component of cell culture to test its effect on rat subventricular zone (SVZ) cells and oligodendrocyte progenitor cells (OPC) proliferation and survival[4]
  • as a model enzyme to track kinetic data and visual detection limits of hydrolysis by hydrolytic enzymes in the two-phases array[5]

Biochem/physiol Actions

Kallikrein active forms are generated by the enzymatic action of trypsin.[1] It is a serine protease that mediates the activation of growth factors and substrates.[2]

Unit Definition

One unit will hydrolyze 1.0 μmole of Nα-benzoyl-L-arginine ethyl ester (BAEE) to Nα-benzoyl-L-arginine and ethanol per min at pH 8.7 at 25°C.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M G Cotenescu et al.
Journal of biotechnology, 76(1), 33-41 (2000-04-28)
A new assay is described that monitors hydrolysis with the concurrent transfer of a solvatochromic dye across an oil-water barrier. Through the appropriate design, this transfer is accompanied by a 10(6) gain in fluorescence. This response can be used to
Gabriel Rosenblum et al.
Journal of the American Chemical Society, 129(44), 13566-13574 (2007-10-13)
Activation of matrix metalloproteinase zymogen (pro-MMP) is a vital homeostatic process, yet its molecular basis remains unresolved. Using stopped-flow X-ray spectroscopy of the active site zinc ion, we determined the temporal sequence of pro-MMP-9 activation catalyzed by tissue kallikrein protease
Generation of alpha- and beta-kallikreins from porcine pancreatic prokallikrein by the action of trypsin.
M Kamada et al.
Chemical & pharmaceutical bulletin, 36(12), 4891-4899 (1988-12-01)
S C Fernando et al.
Genomics, 89(3), 429-438 (2007-01-11)
Kallikreins belong to a family of serine proteases that are widespread throughout living organisms, expressed in diverse tissue-specific patterns, and known to have highly diverse physiological functions. The 15 human and 24 mouse kallikreins have been implicated in pathophysiology of
Ling Liu et al.
Biochemical and biophysical research communications, 407(2), 283-287 (2011-03-08)
Systemic or local delivery of human tissue kallikrein gene (hTK) has been shown to be an effective strategy to alleviate cerebral ischemia/reperfusion (I/R) injury, and tissue kallikrein (TK) administration can suppress glutamate- or acidosis-mediated neurotoxicity in vitro. In the present

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service