Skip to Content
Merck
All Photos(2)

Documents

I9646

Sigma-Aldrich

Interleukin-6 from mouse

IL-6, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture, carrier free

Synonym(s):

mIL-6, IL-6

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

biological source

mouse

Quality Level

recombinant

expressed in E. coli

Assay

≥97% (SDS-PAGE)

form

lyophilized powder

potency

0.02-0.2 ng/mL EC50

quality

endotoxin tested

mol wt

~21.7 kDa

packaging

pkg of 5 and 25 μg

technique(s)

cell culture | mammalian: suitable

impurities

≤1.0EU/mg

color

white

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Il6(16193)

Biochem/physiol Actions

Interleukin-6 (IL-6) is a multifunctional protein originally discovered in the media of cells stimulated with double stranded RNA. IL-6 appears to be directly involved in the responses that occur after infection and injury and may prove to be as important as IL-1 and TNF-α in regulating the acute phase response. IL-6 is reported to be produced by fibroblasts, activated T cells, activated monocytes or macrophages, and endothelial cells. It acts upon a variety of cells, including fibroblasts, myeloid progenitor cells, T cells, B cells and hepatocytes. IL-6 induces multiple effects, as indicated by its numerous synonyms: plasmacytoma growth factor (PCT-GF), interferon-β-2 (IFN-β2), monocyte derived human B cell growth factor, B cell stimulating factor (BSF-2), hepatocyte stimulating factor (HSF), Interleukin Hybridoma/Plasmacytoma-1 (IL-HP1). In addition, IL-6 appears to interact with IL-2 in the proliferation of T lymphocytes. IL-6 also potentiates the proliferative effect of IL-3 on multipotential hematopoietic progenitors.

Physical form

Lyophilized from a sterile filtered solution with no additives.

Analysis Note

The biological activity of recombinant mouse IL-6 was measured in a cell proliferation assay using T1165.85.2.1 cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marrah E Lachowicz-Scroggins et al.
American journal of respiratory cell and molecular biology, 43(6), 652-661 (2010-01-19)
Infection of airway epithelium by rhinovirus is the most common cause of asthma exacerbations. Even in mild asthma, airway epithelium exhibits mucous metaplasia, which increases with increasing severity of the disease. We previously showed that squamous cultures of human airway
Jee Eun Choi et al.
STAR protocols, 3(2), 101389-101389 (2022-05-24)
Metabolic reprogramming is associated with myeloid-derived suppressor cell (MDSC) immunosuppressive function. Here, we outline the process for acquiring MDSCs from human and murine sources for subsequent analysis of fatty acid oxidation, oxidative phosphorylation, and glycolysis using the Seahorse XFe 96
Kimberley C Clark et al.
Cancers, 12(8) (2020-08-07)
In most instances, multiple myeloma (MM) plasma cells (PCs) are reliant on factors made by cells of the bone marrow (BM) stroma for their survival and growth. To date, the nature and cellular composition of the BM tumor microenvironment and
JiSheng Hu et al.
Molecules and cells, 43(8), 749-762 (2020-09-02)
The migration, dedifferentiation, and proliferation of vascular smooth muscle cells (VSMCs) are responsible for intimal hyperplasia, but the mechanism of this process has not been elucidated. WD repeat domain 1 (WDR1) promotes actin-depolymerizing factor (ADF)/cofilin-mediated depolymerization of actin filaments (F-actin).
R P Nordan et al.
Journal of immunology (Baltimore, Md. : 1950), 139(3), 813-817 (1987-08-01)
Plasmacytoma growth factor (PCT-GF), a putative macrophage-derived lymphokine essential for the in vitro viability and proliferation of early generation plasmacytomas, was purified from conditioned medium of the murine macrophage cell line P388D1. The purification of PCT-GF was accomplished by a

Articles

Interleukins, released from leukocytes, include IL-1β playing crucial roles in inflammation, immune responses, and anti-tumor immunity.

Interleukins, released from leukocytes, include IL-1β playing crucial roles in inflammation, immune responses, and anti-tumor immunity.

Interleukins, released from leukocytes, include IL-1β playing crucial roles in inflammation, immune responses, and anti-tumor immunity.

Interleukins, released from leukocytes, include IL-1β playing crucial roles in inflammation, immune responses, and anti-tumor immunity.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service