Skip to Content
Merck
All Photos(1)

Key Documents

I1882

Sigma-Aldrich

Insulin from bovine pancreas

γ-irradiated, BioXtra, suitable for cell culture, potency: ≥20 units/mg (USP units), lyophilized powder

Synonym(s):

Bovine insulin, Cell culture grade insulin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C254H377N65O75S6
CAS Number:
Molecular Weight:
5733.49
EC Number:
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.77
biological source:
bovine
form:
lyophilized powder

biological source

bovine

Quality Level

sterility

γ-irradiated

product line

BioXtra

form

lyophilized powder

potency

≥20 units/mg (USP units)

technique(s)

cell culture | mammalian: suitable

solubility

acetic acid: 2 mg/mL (pH: 2-3)
hydrochloric acid: 2 mg/mL (pH: 2-3)

UniProt accession no.

shipped in

ambient

storage temp.

−20°C

InChI key

IXIBAKNTJSCKJM-BUBXBXGNSA-N

Gene Information

bovine ... INS(280829)

Looking for similar products? Visit Product Comparison Guide

Application

Insulin is used as a growth factor in many mammalian cell culture systems.

Biochem/physiol Actions

Two-chain polypeptide hormone produced by the β-cells of pancreatic islets. Its molecular weight is ~5800 Da. The α and β chains are joined by two interchain disulfide bonds. The α chain contains an intrachain disulfide bond. Insulin regulates the cellular uptake, utilization, and storage of glucose, amino acids, and fatty acids and inhibits the breakdown of glycogen, protein, and fat.

Other Notes

InChI: 1S/C254H377N65O75S6/c1-29-131(24)205(312-192(334)103-256)250(389)317-204(130(22)23)246(385)287-158(74-81-199(344)345)216(355)281-155(70-77-188(260)330)220(359)307-183-116-399-400-117-184-242(381)305-177(110-321)238(377)293-161(87-122(6)7)223(362)294-167(94-139-52-60-145(324)61-53-139)226(365)282-153(68-75-186(258)328)217(356)289-160(86-121(4)5)221(360)284-157(73-80-198(342)343)219(358)301-173(100-189(261)331)233(372)297-169(96-141-56-64-147(326)65-57-141)229(368)308-182(241(380)303-175(253(393)394)102-191(263)333)115-398-396-113-180(212(351)270-106-193(335)277-152(71-78-196(338)339)215(354)280-150(50-41-83-268-254(264)265)210(349)269-107-194(336)278-165(92-137-45-35-31-36-46-137)225(364)296-166(93-138-47-37-32-38-48-138)228(367)298-170(97-142-58-66-148(327)67-59-142)236(375)318-206(135(28)323)251(390)319-84-42-51-185(319)244(383)285-151(49-39-40-82-255)213(352)276-134(27)252(391)392)310-248(387)202(128(18)19)315-234(373)163(89-124(10)11)291-227(366)168(95-140-54-62-146(325)63-55-140)295-222(361)159(85-120(2)3)288-207(346)132(25)274-214(353)156(72-79-197(340)341)286-245(384)201(127(16)17)314-235(374)164(90-125(12)13)292-231(370)172(99-144-105-267-119-273-144)300-237(376)176(109-320)279-195(337)108-271-211(350)179(112-395-397-114-181(309-243(183)382)240(379)275-133(26)208(347)304-178(111-322)239(378)316-203(129(20)21)249(388)311-184)306-224(363)162(88-123(8)9)290-230(369)171(98-143-104-266-118-272-143)299-218(357)154(69-76-187(259)329)283-232(371)174(101-190(262)332)302-247(386)200(126(14)15)313-209(348)149(257)91-136-43-33-30-34-44-136/h30-38,43-48,52-67,104-105,118-135,149-185,200-206,320-327H,29,39-42,49-51,68-103,106-117,255-257H2,1-28H3,(H2,258,328)(H2,259,329)(H2,260,330)(H2,261,331)(H2,262,332)(H2,263,333)(H,266,272)(H,267,273)(H,269,349)(H,270,351)(H,271,350)(H,274,353)(H,275,379)(H,276,352)(H,277,335)(H,278,336)(H,279,337)(H,280,354)(H,281,355)(H,282,365)(H,283,371)(H,284,360)(H,285,383)(H,286,384)(H,287,385)(H,288,346)(H,289,356)(H,290,369)(H,291,366)(H,292,370)(H,293,377)(H,294,362)(H,295,361)(H,296,364)(H,297,372)(H,298,367)(H,299,357)(H,300,376)(H,301,358)(H,302,386)(H,303,380)(H,304,347)(H,305,381)(H,306,363)(H,307,359)(H,308,368)(H,309,382)(H,310,387)(H,311,388)(H,312,334)(H,313,348)(H,314,374)(H,315,373)(H,316,378)(H,317,389)(H,318,375)(H,338,339)(H,340,341)(H,342,343)(H,344,345)(H,391,392)(H,393,394)(H4,264,265,268)/t131-,132-,133-,134-,135+,149-,150-,151-,152-,153-,154-,155-,156-,157-,158-,159-,160-,161-,162-,163-,164-,165-,166-,167-,168-,169-,170-,171-,172-,173-,174-,175-,176-,177-,178-,179-,180-,181-,182-,183-,184-,185-,200-,201-,202-,203-,204-,205-,206-/m0/s1

Physical form

powder-0 °C; solution stable 2-8 °C, 1 year

Reconstitution

To prepare 10 mg/ml stock solution, add 10 ml of acidified H2O (pH ≤2) - prepared by addition of glacial acetic acid (approx. 0.1 ml).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kevin M Dean et al.
Scientific reports, 7(1), 9332-9332 (2017-08-26)
We introduce a concept that enables parallelized three-dimensional imaging throughout large volumes with isotropic 300-350 nm resolution. By staggering high aspect ratio illumination beams laterally and axially within the depth of focus of a digitally scanned light-sheet fluorescence microscope (LSFM), multiple
Kristina Attoff et al.
PloS one, 12(12), e0190066-e0190066 (2017-12-21)
Despite its high relevance, developmental neurotoxicity (DNT) is one of the least studied forms of toxicity. Current guidelines for DNT testing are based on in vivo testing and they require extensive resources. Transcriptomic approaches using relevant in vitro models have
Marc Hafner et al.
Cell chemical biology, 26(8), 1067-1080 (2019-06-11)
The target profiles of many drugs are established early in their development and are not systematically revisited at the time of FDA approval. Thus, it is often unclear whether therapeutics with the same nominal targets but different chemical structures are
Sarfraz Ahmad et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 17(6), 1101-1108 (2009-04-02)
Prostate stem cell antigen (PSCA) is a cell surface antigen expressed in normal human prostate and over expressed in prostate cancer. Elevated levels of PSCA protein in prostate cancer correlate with increased tumor stage/grade, with androgen independence and have higher
Ian S Zagon et al.
Archives of ophthalmology (Chicago, Ill. : 1960), 127(11), 1468-1473 (2009-11-11)
To determine if topical application of naltrexone hydrochloride (NTX), an opioid antagonist, restores tear production and corneal sensation in rats with diabetes mellitus. Type 1 diabetes was induced with streptozotocin in rats. Tear production was measured by the Schirmer test

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service