Skip to Content
Merck
All Photos(2)

Key Documents

HS1420A

Weigh Boat

size small, square white polystyrene

Synonym(s):

weigh boat, weigh dish, weighing dish, weighing pan

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121812
NACRES:
NB.71

material

square white polystyrene

size

small

packaging

pack of 500 ea

manufacturer/tradename

Heathrow Scientific HS1420A

L × W × H

46 mm × 46 mm × 8 mm

Looking for similar products? Visit Product Comparison Guide

General description

Flexible polystyrene boats feature smooth surfaces to provide accurate pour-outs with minimal sample loss.
The Standard Weighing Boat is flexible and features smooth surfaces to provide accurate pour-outs with minimal sample loss.

Features and Benefits

  • Boat is shallow with a wide, flat bottom to resist tipping
  • Rounded corners simplify transfer
  • Color contrast to ensure sample is completely removed
  • Biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases
  • Temperature range -10 to 70°C (14 to 158°F)
  • Can be used with microwave moisture analyzers
  • FDA/food-grade compliant
  • Disposable
  • Manufactured and packaged in a clean room
  • biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases.
  • temperature range -10 to 70C
  • disposable
  • shallow with a wide, flat bottom to resist tipping
  • rounded corners simplify transfer


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service