Skip to Content
Merck
All Photos(5)

Key Documents

HPA017743

Sigma-Aldrich

Anti-PRSS16 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Serine protease 16, Anti-Thymus-specific serine protease precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

HSTPYCGLRRAVQIVLHSLGQKCLSFSRAETVAQLRSTEPQLSGVGDRQWLYQTCTEFGFYVTCENPRCPFSQLPALPSQLDLCEQVFGLSALSVAQAVAQTNSYYGG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRSS16(10279)

General description

Thymus-specific serine protease (PRSS16) is present in epithelial cells of the thymic cortex. The gene encoding this protein is localized to chromosome 6p21.3-p22.

Immunogen

Thymus-specific serine protease precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Thymus-specific serine protease (PRSS16) may play a role in immune function and positive selection of T cells. It has been shown to be associated with schizophrenia. PRSS16 may aid in the development of the embryogenic thymus and regulate events during thymopoiesis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70802

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saijai Cheunsuk et al.
Journal of autoimmunity, 18(4), 311-316 (2002-07-30)
PRSS16 is a serine protease specifically expressed by epithelial cells in the thymic cortex. The human gene is encoded on 6p21.3-p22 where recent linkage analysis has identified an association with insulin dependent diabetes mellitus (IDDM) susceptibility independent of HLA-DR3. To
C Luther et al.
Genes and immunity, 6(1), 1-7 (2004-12-14)
The putative serine protease PRSS16 is abundantly expressed in the thymic cortex and the gene is encoded within the HLA I complex. Although its function is not yet defined, the very restricted expression points to a role in T-cell development
M K Viken et al.
Genes and immunity, 10(4), 323-333 (2009-03-20)
The high-risk human leukocyte antigen (HLA)-DRB1, DQA1 and DQB1 alleles cannot explain the entire type 1 diabetes (T1D) association observed within the extended major histocompatibility complex. We have earlier identified an association with D6S2223, located 2.3 Mb telomeric of HLA-A
Matthew J Girgenti et al.
PloS one, 7(2), e32404-e32404 (2012-03-03)
ZNF804a was identified by a genome-wide association study (GWAS) in which a single nucleotide polymorphism (SNP rs1344706) in ZNF804a reached genome-wide statistical significance for association with a combined diagnosis of schizophrenia (SZ) and bipolar disorder. Although the molecular function of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service