Skip to Content
Merck
All Photos(4)

Key Documents

HPA004145

Sigma-Aldrich

Anti-AHNAK2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-AL512802.2, Anti-Putative uncharacterized protein ENSP00000381351 fragment

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AHNAK2(113146)

General description

AHNAK nucleoprotein 2 is a large polypeptide with 600kDa molecular weight. Its giant propeller-like structure is composed of interconnected antiparallel β-strands with PDZ domain within its N-terminal, nonrepeating domain. It is localized at the interphase region of nuclei as a differentiation-related protein.

Immunogen

AHNAK nucleoprotein 2 recombinant protein epitope signature tag (PrEST)

Sequence
APRAKLDSAQLEGDLSLADKDVTAKDSKFKMPKFKMPSFGVSAPGKSIEASVHVSAPKVEADVSLPSMQGDLKTTDLSIQPHSADLTVQARQVDMKLLEGHVPEEAGLKGHLPKVQMPSFKMPKVDLKG

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Depending on the presence of arachidonic acid, AHNAK2 binds to the phospholipase C-γ (PLC-γ). It activates PLC by binding to it, which finally generates diacylglycerol and inositol trisphosphate. These two by products of phosphoinositide hydrolysis are produced by ligand-stimulated receptors or G protein activation of PLCs. It also functions as a tumour-related phosphoprotein in a wide range of physiological activities.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70636

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hannelore Haase
Cardiovascular research, 73(1), 19-25 (2006-10-19)
Ahnak, originally identified as a giant, tumour-related phosphoprotein, has emerged as an important signalling molecule in a wide range of physiological activities. In this article, current knowledge will be reviewed that places ahnak into the context of cardiac L-type Ca2+
Di Lu et al.
Oncotarget, 8(19), 31775-31784 (2017-04-21)
AHNAK nucleoprotein 2 (AHNAK2) belongs to the AHNAK protein family. The studies of AHNAK2 are limited. A recent study reported that AHNAK2 might be a biomarker for pancreatic ductal adenocarcinoma (PDAC); however, tissue-based experiments have not been conducted. The aim
F Sekiya et al.
The Journal of biological chemistry, 274(20), 13900-13907 (1999-05-13)
We have recently shown that phospholipase C-gamma (PLC-gamma) is activated by tau, a neuronal cell-specific microtubule-associated protein, in the presence of arachidonic acid. We now report that non-neuronal tissues also contain a protein that can activate PLC-gamma in the presence
Dai Koguchi et al.
Cancers, 13(8) (2021-05-01)
Data regarding expression levels of AHNAK2 in bladder cancer (BCa) have been very scarce. We retrospectively reviewed clinical data including clinicopathological features in 120 patients who underwent radical cystectomy (RC) for BCa. The expression levels of AHNAK2 in the specimens
Shusen Zhang et al.
BioMed research international, 2020, 8571932-8571932 (2020-09-10)
Lung adenocarcinoma (LUAD), as the main subtype of lung cancer, is one of the common causes of cancer-related deaths worldwide. The AHNAK family is correlated with cell structure and migration, cardiac calcium channel signaling, and tumor metastasis. Previous studies showed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service