Skip to Content
Merck
All Photos(1)

Key Documents

H5884

Sigma-Aldrich

22(S)-Hydroxycholesterol

Synonym(s):

22β-Hydroxycholesterol, 5-Cholestene-3β,22(S)-diol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H46O2
CAS Number:
Molecular Weight:
402.65
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (TLC)

Quality Level

form

powder

shipped in

ambient

storage temp.

room temp

SMILES string

[H][C@@]12[C@]([C@](CC[C@H](O)C3)(C)C3=CC2)([H])CC[C@@]4(C)[C@@]1([H])CC[C@]4([H])[C@]([H])(C)[C@@H](O)CCC(C)C

InChI

1S/C27H46O2/c1-17(2)6-11-25(29)18(3)22-9-10-23-21-8-7-19-16-20(28)12-14-26(19,4)24(21)13-15-27(22,23)5/h7,17-18,20-25,28-29H,6,8-16H2,1-5H3/t18-,20-,21-,22+,23-,24-,25-,26-,27+/m0/s1

InChI key

RZPAXNJLEKLXNO-QUOSNDFLSA-N

Application

Human hepatoma cells were treated with 22(S)-Hydroxycholesterol to study cholesterol modulation by liver X receptor-mediated gene regulation.

Biochem/physiol Actions

22(S)-Hydroxycholesterol modulates the lipid and glucose metabolism by acting on liver X receptors. It has the potential to reduce lipogenesis and the accumulation of lipid in hepatocytes, myotubes and non-adipose tissue.

Preparation Note

22(S)-Hydroxycholesterol yields clear, colorless solution in chloroform at 20 mg/ml.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

E T Kase et al.
Diabetologia, 50(10), 2171-2180 (2007-07-31)
Liver X receptors (LXRs) play important roles in lipid and carbohydrate metabolism. The purpose of the present study was to evaluate effects of the endogenous LXR agonist 22-R-hydroxycholesterol (22-R-HC) and its stereoisomer 22-S-hydroxycholesterol (22-S-HC), in comparison with the synthetic agonist
Eili Tranheim Kase et al.
Biochimica et biophysica acta, 1761(12), 1515-1522 (2006-10-24)
The nuclear liver X receptors (LXRalpha and beta) are regulators of lipid and cholesterol metabolism. Oxysterols are known LXR ligands, but the functional role of hydroxycholesterols is at present unknown. In human myotubes, chronic exposure to the LXR ligand T0901317
K Hanley et al.
The Journal of investigative dermatology, 114(3), 545-553 (2000-02-26)
Ligands and activators of the nuclear hormone receptor superfamily are important in the regulation of epidermal development and differentiation. Previously, we showed that naturally occurring fatty acids, as well as synthetic ligands for the peroxisome proliferator-activated receptor, induce keratinocyte differentiation
B A Laffitte et al.
Molecular and cellular biology, 21(22), 7558-7568 (2001-10-18)
Previous work has implicated the nuclear receptors liver X receptor alpha (LXR alpha) and LXR beta in the regulation of macrophage gene expression in response to oxidized lipids. Macrophage lipid loading leads to ligand activation of LXRs and to induction
Bassam Haidar et al.
The Journal of biological chemistry, 279(11), 9963-9969 (2004-01-01)
It has been suggested that the signal transduction pathway initiated by apoA-I activates key proteins involved in cellular lipid efflux. We investigated apoA-I-mediated cAMP signaling in cultured human fibroblasts induced with (22R)-hydroxycholesterol and 9-cis-retinoic acid (stimulated cells). Treatment of stimulated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service