Skip to Content
Merck
All Photos(1)

Documents

GE28-9782-44

HiScreen Ni Fast Flow

Cytiva 28-9782-44

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
13111000
NACRES:
NA.56

packaging

pkg of 1 ea

manufacturer/tradename

Cytiva 28-9782-44

parameter

1.5 bar (22 psi) (Over the Packed Bed During Operation)
22 psi

bed size

7.7 mm × 100 mm

bed volume

4.7 mL

column I.D.

7.7 mm

matrix

6% cross-linked agarose

particle size

45-165 μm

average diameter

90 μm

cleaning

2-14(Ni2+-stripped medium.)

working range

3-12(Ni2+-stripped medium.)

capacity

~40 mg binding capacity(histidine-tagged protein)

suitability

suitable for bioprocess medium

General description

HiScreen columns prepacked with Ni Sepharose 6 Fast FLow for method optimization and parameter screening for purification of histidine-tagged proteins with Immobilized Metal ion Affinity Chromatography (IMAC).

Application

HiScreen columns are part of the process development platform available from Cytiva. The columns are prepacked with a range of BioProcess chromatography media (resin) and designed for method optimization and parameter screening. HiScreen columns have small bed volumes (4.7 ml), reducing the cost of sample and buffer consumption. The chromatography media used in HiScreen columns are also available in other prepacked formats and as bulk packs, for all scales of work from development and pilot studies to routine production.

HiScreen Ni FF is prepacked with the BioProcess chromatography medium Ni Sepharose 6 Fast FLow which has high binding capacities for
histidine-tagged Proteins and minimal Ni2+ leakage.

Features and Benefits

  • High binding capacity, approx. 40 mg/mL medium and negligible leakage of Ni2+.
  • Prepacked, ready-to-use columns for convenient process development.
  • Excellent choice for method optimization and parameter screening due to the 10 cm bed height.
  • Easy connection in series to achieve 20 cm bed height.
  • Small bed volume gives fast results and minimal sample/buffer consumption
  • Reproducible results, scalable to BioProcess columns packed with the same media using the same linear fluid velocity

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

HiScreen is a trademark of Cytiva
Sepharose is a trademark of Cytiva

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page shows the characteristics of Ni Sepharose, Ni Sepharose excel, TALON Superflow, and uncharged IMAC Sepharose products from Cytiva.

This page shows the characteristics of Ni Sepharose, Ni Sepharose excel, TALON Superflow, and uncharged IMAC Sepharose products from Cytiva.

This page shows the characteristics of Ni Sepharose, Ni Sepharose excel, TALON Superflow, and uncharged IMAC Sepharose products from Cytiva.

This page shows the characteristics of Ni Sepharose, Ni Sepharose excel, TALON Superflow, and uncharged IMAC Sepharose products from Cytiva.

See All

Protocols

Ni Sepharose 6 Fast Flow purifies histidine-tagged proteins efficiently, offering high cross-linked agarose beads with Ni2+ ions.

Ni Sepharose 6 Fast Flow purifies histidine-tagged proteins efficiently, offering high cross-linked agarose beads with Ni2+ ions.

Ni Sepharose 6 Fast Flow purifies histidine-tagged proteins efficiently, offering high cross-linked agarose beads with Ni2+ ions.

Ni Sepharose 6 Fast Flow purifies histidine-tagged proteins efficiently, offering high cross-linked agarose beads with Ni2+ ions.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service