Skip to Content
Merck
All Photos(1)

Documents

G8541

Sigma-Aldrich

Ala-Gln

200 mM, solution, sterile-filtered, Biotechnology Performance Certified

Synonym(s):

Alanyl-glutamine, Glutamine-S

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15N3O4
CAS Number:
Molecular Weight:
217.22
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.75

grade

Biotechnology Performance Certified

sterility

sterile-filtered

form

solution

concentration

200 mM

impurities

endotoxin, tested

storage temp.

2-8°C

SMILES string

NC(CC[C@H](NC([C@@H](N)C)=O)C(O)=O)=O

InChI

1S/C8H15N3O4/c1-4(9)7(13)11-5(8(14)15)2-3-6(10)12/h4-5H,2-3,9H2,1H3,(H2,10,12)(H,11,13)(H,14,15)/t4-,5-/m0/s1

InChI key

HJCMDXDYPOUFDY-WHFBIAKZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ala-Gln supplement provides a stable form of L-glutamine while in solution.

Application

Ala-Gln has been used as a supplement in Dulbecco′s Modified Eagle Medium (DMEM) media-rich in glucose and pyruvate to culture luciferase-expressing MDA-MB-231-luc tumor cells to measure the bioluminescence signals of cells leaking during intra-bone marrow transplantation (IBMT) from dissected long bones. It has also been used as a DMEM - high glucose media to culture human cell lines.

Other Notes

Contains the dipeptide L-Alanyl-L-glutamine. 200 mM supplied in 0.85% NaCl.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daren K Heyland et al.
JPEN. Journal of parenteral and enteral nutrition, 39(4), 401-409 (2014-05-08)
The recent large randomized controlled trial of glutamine and antioxidant supplementation suggested that high-dose glutamine is associated with increased mortality in critically ill patients with multiorgan failure. The objectives of the present analyses were to reevaluate the effect of supplementation
Dieter Fink et al.
PloS one, 14(11), e0224576-e0224576 (2019-11-08)
Intra-bone marrow transplantation (IBMT) has been adapted for mouse models to improve the seeding efficiency of transplanted hematopoietic stem and progenitor cells. Commonly used injection volumes for IBMT into the tibia differ between 10 and 40 μL even though considerable
S Taavitsainen et al.
Nature communications, 12(1), 5307-5307 (2021-09-08)
Prostate cancer is heterogeneous and patients would benefit from methods that stratify those who are likely to respond to systemic therapy. Here, we employ single-cell assays for transposase-accessible chromatin (ATAC) and RNA sequencing in models of early treatment response and
Marek Bartosovic et al.
Nature biotechnology, 39(7), 825-835 (2021-04-14)
In contrast to single-cell approaches for measuring gene expression and DNA accessibility, single-cell methods for analyzing histone modifications are limited by low sensitivity and throughput. Here, we combine the CUT&Tag technology, developed to measure bulk histone modifications, with droplet-based single-cell
Shaoquan Lin et al.
Nature chemistry, 13(1), 47-55 (2020-12-24)
Polyether ionophores are complex natural products capable of transporting cations across biological membranes. Many polyether ionophores possess potent antimicrobial activity and a few selected compounds have the ability to target aggressive cancer cells. Nevertheless, ionophore function is believed to be

Articles

Importance and uses of glutamine in hybridoma and mammalian cell culture

Importance and uses of glutamine in hybridoma and mammalian cell culture

Importance and uses of glutamine in hybridoma and mammalian cell culture

Importance and uses of glutamine in hybridoma and mammalian cell culture

See All

Related Content

Monitor barrier formation using colon PDOs, iPSC-derived colon organoids, Millicell® cell culture inserts, and the Millicell® ERS. 3.0.

Monitor barrier formation using colon PDOs, iPSC-derived colon organoids, Millicell® cell culture inserts, and the Millicell® ERS. 3.0.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service