Skip to Content
Merck
All Photos(1)

Key Documents

G8168

Sigma-Aldrich

G418 disulfate salt

solution, non-animal origin, suitable for cell culture, BioReagent

Synonym(s):

Antibiotic G418

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H40N4O10 · 2H2SO4
CAS Number:
Molecular Weight:
692.71
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.76

Product Name

G 418 disulfate salt solution, 50 mg/mL in H2O, 0.1 μm filtered, BioReagent, suitable for cell culture

biological source

Micromonospora echinospora

Quality Level

sterility

0.1 μm filtered

product line

BioReagent

form

solution

potency

50-60 mg per mL

mol wt

692.7

packaging

pkg of 10 and 100 mL

concentration

50 mg/mL in H2O

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

color

colorless

solubility

H2O: 50 mg/mL, clear

antibiotic activity spectrum

parasites

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

OS(O)(=O)=O.OS(O)(=O)=O.CN[C@H]1[C@H](O)[C@@H](OC[C@@]1(C)O)O[C@H]2[C@H](N)C[C@H](N)[C@@H](O[C@H]3O[C@H]([C@H](C)O)[C@@H](O)[C@H](O)[C@H]3N)[C@@H]2O

InChI

1S/C20H40N4O10.2H2O4S/c1-6(25)14-11(27)10(26)9(23)18(32-14)33-15-7(21)4-8(22)16(12(15)28)34-19-13(29)17(24-3)20(2,30)5-31-19;2*1-5(2,3)4/h6-19,24-30H,4-5,21-23H2,1-3H3;2*(H2,1,2,3,4)/t6?,7-,8+,9+,10+,11-,12-,13-,14+,15+,16-,17-,18+,19-,20+;;/m0../s1

InChI key

UHEPSJJJMTWUCP-NKCAIAFTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: aminoglycoside

Application

G418 disulfate is an aminoglycoside antibiotic similar in structure to gentamicin used for selection and maintenance of prokaryotic and eukaryotic cells. Optimal concentrations for selection and maintenance vary by cell lines. Bacteria and algae require 5 μg/mL or less while animal cells may require 300-500 μg/mL.

Biochem/physiol Actions

Mode of Action: G418 blocks polypeptide synthesis and protein elongation by inhibiting synthesis at the 70S and 80S ribosomes.

Antimicrobial Spectrum: G418 selects for cells stably transfected with an iNOS promoter construct and neomycin resistance gene and shows activity against protozoa and helminthes.

Caution

The product is stable for two years when stored at 2-8°C.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jieru Li et al.
Nature structural & molecular biology, 27(11), 1032-1040 (2020-09-23)
Transcription activation by distal enhancers is essential for cell-fate specification and maintenance of cellular identities. How long-range gene regulation is physically achieved, especially within complex regulatory landscapes of non-binary enhancer-promoter configurations, remains elusive. Recent nanoscopy advances have quantitatively linked promoter
Lene Halkjær et al.
Frontiers in immunology, 11, 774-774 (2020-05-21)
We aimed at establishing a sensitive and robust assay for estimation of systemic complement activation at complement component C3 level in mouse and human plasma samples. In order to capture the activation products iC3b and C3dg in a specific and
Kelvin Kai-Wan Hui et al.
Journal of cellular and molecular medicine, 23(3), 1784-1797 (2018-12-15)
Aberrant regulation of programmed cell death (PCD) has been tied to an array of human pathologies ranging from cancers to autoimmune disorders to diverse forms of neurodegeneration. Pharmacologic modulation of PCD signalling is therefore of central interest to a number
Alaullah Sheikh et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(46), 29055-29062 (2020-11-04)
The enterotoxigenic Escherichia coli (ETEC) are among the most common causes of diarrheal illness and death due to diarrhea among young children in low-/middle-income countries (LMICs). ETEC have also been associated with important sequelae including malnutrition and stunting, placing children
Frida C Mohlin et al.
Molecular immunology, 65(2), 367-376 (2015-03-04)
Atypical hemolytic uremic syndrome (aHUS) is a disease of complement dysregulation, characterized by hemolytic anemia, thrombocytopenia and acute renal failure. Mutations in complement inhibitors are major risk factors for development of aHUS. The three aHUS patients reported in this study

Articles

Lentiviral vector systems prioritize safety features, with design precautions preventing replication. Good handling practices are essential for use.

Lentiviral vector systems prioritize safety features, with design precautions preventing replication. Good handling practices are essential for use.

Lentiviral vector systems prioritize safety features, with design precautions preventing replication. Good handling practices are essential for use.

Lentiviral vector systems prioritize safety features, with design precautions preventing replication. Good handling practices are essential for use.

See All

Protocols

Lentivirus versions of genome modification technologies support successful CRISPR, RNAi, and ORF experiments.

Lentivirus versions of genome modification technologies support successful CRISPR, RNAi, and ORF experiments.

Lentivirus versions of genome modification technologies support successful CRISPR, RNAi, and ORF experiments.

Lentivirus versions of genome modification technologies support successful CRISPR, RNAi, and ORF experiments.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service