Skip to Content
Merck
All Photos(1)

Key Documents

G8046

Sigma-Aldrich

Anti-phospho-G3BP (pSer149) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-G3BP1, Anti-GAP SH3 domain-binding protein, Anti-HDH-VIII, Anti-RAS-GTPase-activating protein SH3-domain binding protein 1, Anti-human DNA helicase VIII

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~68 kDa

species reactivity

human, rat (predicted), mouse (predicted)

concentration

~1.0 mg/mL

technique(s)

western blot: 5-10 μg/mL using whole extract of human K562 and HeLa cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer149)

Gene Information

human ... G3BP1(10146)
mouse ... G3bp1(27041)
rat ... G3bp1(171092)

Related Categories

General description

G3BP (Ras-GTPase-activating protein SH3 domain binding protein 1) is a 68kDa, single-strand-specific endoribonuclease that is phosphorylation-dependent.

Application

Anti-phospho-G3BP (pSer149) antibody produced in rabbit is suitable for western blotting at a working concentration of 5-10μg/mL using whole extract of human K562 and HeLa cells.

Biochem/physiol Actions

G3BP (Ras-GTPase-activating protein SH3 domain binding protein 1) exclusively cleaves between cytosine and adenine (CA). In dividing cells, G3BP interacts with RasGAP, linking between a RasGAP-mediated signaling pathway and RNA turnover. It is a RNA-binding protein that contains a carboxyl-terminal RNA binding domain, the RRM-type domain. It also has an amino-terminal domain homologous to nuclear transporter factor 2 (NTF2), and a central domain rich in acidic residues. The RRM domain is involved in the binding of G3BP to specific RNA sequences. A Ser149 residue is present 20 amino acids C terminal to the NTF2-like domain. Phosphorylation of this residue is crucial in mediating protein-protein interactions and in controlling the subcellular localization of G3BP. G3BP facilitates the assembly of stress granules that are invoved in the regulation of mRNA metabolism during stress. Dephosphorylation of Ser149 results in oligomerization and SG assembly. Overexpression of G3BP is observed in malignant tumors such as lung cancer, colon cancer, gastric cancer, and breast cancer. The level of G3BP expression in breast cancer specimens is positively correlated to the presence of lymph node metastasis.

Target description

G3BP (pSer149) encodes one of the DNA-unwinding enzymes which prefers partially unwound 3′-tailed substrates and can also unwind partial RNA/DNA and RNA/RNA duplexes in an ATP-dependent fashion.

Physical form

Solution 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marc D Panas et al.
The Journal of cell biology, 218(7), 2425-2432 (2019-06-07)
Tourrière et al. (2013. J. Cell Biol. https://doi.org/10.1083/jcb.200212128) reported that G3BP1-S149 dephosphorylation promotes stress granule formation. We show that constructs used to establish this conclusion contain additional mutations causing these phenotypes, and that S149 phosphorylation status does not change upon
Paul Anderson et al.
The Journal of cell biology, 172(6), 803-808 (2006-03-08)
Cytoplasmic RNA granules in germ cells (polar and germinal granules), somatic cells (stress granules and processing bodies), and neurons (neuronal granules) have emerged as important players in the posttranscriptional regulation of gene expression. RNA granules contain various ribosomal subunits, translation
Monika Jedrusik-Bode et al.
Journal of cell science, 126(Pt 22), 5166-5177 (2013-09-10)
SIRT6 is a NAD(+)-dependent deacetylase that modulates chromatin structure and safeguards genomic stability. Until now, SIRT6 has been assigned to the nucleus and only nuclear targets of SIRT6 are known. Here, we demonstrate that in response to stress, C. elegans
H Tourrière et al.
Molecular and cellular biology, 21(22), 7747-7760 (2001-10-18)
Mitogen activation of mRNA decay pathways likely involves specific endoribonucleases, such as G3BP, a phosphorylation-dependent endoribonuclease that associates with RasGAP in dividing but not quiescent cells. G3BP exclusively cleaves between cytosine and adenine (CA) after a specific interaction with RNA
Hui-Zhong Zhang et al.
World journal of gastroenterology, 13(30), 4126-4130 (2007-08-19)
To investigate the expression and significance of G3BP and RhoC proteins in esophageal squamous carcinoma (ESC). The expression of G3BP and Rhoc proteins in 80 cases of ESC was detected by immunohistochemistry. The relationship was studied between the expression of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service