G7276
Gibberellin A4
suitable for plant cell culture, BioReagent, ≥90%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C19H24O5
CAS Number:
Molecular Weight:
332.39
EC Number:
MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:
NACRES:
NA.72
Assay:
≥90%
Recommended Products
product line
BioReagent
Quality Level
Assay
≥90%
technique(s)
cell culture | plant: suitable
impurities
≤5% gibberellin A7
application(s)
agriculture
storage temp.
−20°C
SMILES string
[H][C@@]12CC[C@@H]3C[C@]1(CC3=C)[C@@H](C(O)=O)[C@]4([H])[C@@]5(C)[C@@H](O)CC[C@@]24OC5=O
InChI
1S/C19H24O5/c1-9-7-18-8-10(9)3-4-11(18)19-6-5-12(20)17(2,16(23)24-19)14(19)13(18)15(21)22/h10-14,20H,1,3-8H2,2H3,(H,21,22)/t10?,11-,12+,13-,14-,17-,18+,19-/m1/s1
InChI key
RSQSQJNRHICNNH-AMNRFOROSA-N
Application
Gibberellin A4 has been used as a supplement in Murashige and Skoog (MS) media for enhancing plant growth.
Biochem/physiol Actions
Gibberillic acids (GA) are important plant growth hormones that promote plant cell growth and elongation. Gebberillins promote rapid stem and root growth, induce mitotic division and initiate (break dormancy) and increase seed germination rates. The gibberellins are also involved in processes such as gravitropism, tensioning and floral display.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Annalisa Rizza et al.
Proceedings of the National Academy of Sciences of the United States of America, 118(8) (2021-02-20)
Control over cell growth by mobile regulators underlies much of eukaryotic morphogenesis. In plant roots, cell division and elongation are separated into distinct longitudinal zones and both division and elongation are influenced by the growth regulatory hormone gibberellin (GA). Previously
Dana R MacGregor et al.
The New phytologist, 205(2), 642-652 (2014-11-21)
Environmental changes during seed production are important drivers of lot-to-lot variation in seed behaviour and enable wild species to time their life history with seasonal cues. Temperature during seed set is the dominant environmental signal determining the depth of primary
Seed production temperature regulation of primary dormancy occurs through control of seed coat phenylpropanoid metabolism
MacGregor DR, et al.
The New phytologist, 205(2), 642-652 (2015)
Katie Abley et al.
eLife, 10 (2021-06-02)
Genetically identical plants growing in the same conditions can display heterogeneous phenotypes. Here we use Arabidopsis seed germination time as a model system to examine phenotypic variability and its underlying mechanisms. We show extensive variation in seed germination time variability
Qing Sang et al.
The Plant cell, 32(5), 1479-1500 (2020-03-07)
Several pathways conferring environmental flowering responses in Arabidopsis (Arabidopsis thaliana) converge on developmental processes that mediate the floral transition in the shoot apical meristem. Many characterized mutations disrupt these environmental responses, but downstream developmental processes have been more refractory to
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service