Skip to Content
Merck
All Photos(2)

Key Documents

F3667

Sigma-Aldrich

Fibronectin Adhesion-promoting Peptide

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H74N16O10
CAS Number:
Molecular Weight:
1023.19
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

biological source

synthetic (organic)

Assay

≥95% (HPLC)

form

powder

composition

Peptide content, ~70%

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](C)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H]1CCCN1C(=O)[C@@H]2CCCN2C(=O)[C@H](CCC(N)=O)NC(=O)[C@@H](N)Cc3c[nH]c4ccccc34)C(O)=O

InChI

1S/C47H74N16O10/c1-4-25(2)37(45(72)73)61-41(68)32(14-8-20-55-47(52)53)58-38(65)26(3)57-40(67)31(13-7-19-54-46(50)51)59-42(69)34-15-9-21-62(34)44(71)35-16-10-22-63(35)43(70)33(17-18-36(49)64)60-39(66)29(48)23-27-24-56-30-12-6-5-11-28(27)30/h5-6,11-12,24-26,29,31-35,37,56H,4,7-10,13-23,48H2,1-3H3,(H2,49,64)(H,57,67)(H,58,65)(H,59,69)(H,60,66)(H,61,68)(H,72,73)(H4,50,51,54)(H4,52,53,55)/t25-,26-,29-,31-,32-,33-,34-,35-,37-/m0/s1

InChI key

OVXIMRGEBNSORH-PJRZOMOUSA-N

Amino Acid Sequence

Trp-Gln-Pro-Pro-Arg-Ala-Arg-Ile

Other Notes

Sequence found in the carboxy-terminal heparin-binding domain of fibronectin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Woods et al.
Molecular biology of the cell, 4(6), 605-613 (1993-06-01)
Cell adhesion to extracellular matrix molecules such as fibronectin involves complex transmembrane signaling processes. Attachment and spreading of primary fibroblasts can be promoted by interactions of cell surface integrins with RGD-containing fragments of fibronectin, but the further process of focal

Articles

Fibronectin (FN) plays crucial roles in extracellular matrix fibril assembly and cellular interactions.

Fibronectin (FN) plays crucial roles in extracellular matrix fibril assembly and cellular interactions.

Fibronectin (FN) plays crucial roles in extracellular matrix fibril assembly and cellular interactions.

Fibronectin (FN) plays crucial roles in extracellular matrix fibril assembly and cellular interactions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service