Skip to Content
Merck
All Photos(1)

Documents

E0287

Sigma-Aldrich

Carboxylesterase 1 isoform b human

recombinant, expressed in baculovirus infected BTI insect cells

Synonym(s):

Carboxylesterase 1 human, carboxylesterase, esterase

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
EC Number:
UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in baculovirus infected BTI insect cells

Quality Level

form

liquid

specific activity

≥500 units/mg protein

concentration

5 mg/mL

weight

(0.5 ml)

shipped in

dry ice

storage temp.

−70°C

Looking for similar products? Visit Product Comparison Guide

General description

Carboxylesterase (CE) belongs to serine hydrolase family. It is widely present in the human body. Five isoforms of carboxylesterase is present in humans. It is encoded by CES1 gene mapped to 16q22.2 in human chromosome.
Carboxylesterase 1 is a member of a large multigene carboxylesterase family. These enzymes are responsible for the hydrolysis of ester- and amide-bond-containing drugs such as cocaine and heroin. They also hydrolyze long-chain fatty acid esters and thioesters. This enzyme is known to hydrolyze aromatic and aliphatic esters and is necessary for cellular cholesterol esterification. It may also play a role in detoxification in the lung and/or protection of the central nervous system from ester or amide compounds.

Application

Carboxylesterase 1 isoform b human has been used in the reaction to convert Amplex red to resorufin in horse radish peroxidase and oxygen independent manner. It has also been used in the development of an emission ratiometric two-photon probe (SE1) for quantitation in situ.
Delivers high catalytic activity, ideal for robust high-throughput screening assays including drug-drug interaction studies, and pharmacokinetic studies for evaluating pro-drugs and non-CYP pathways of elimination.

Biochem/physiol Actions

Carboxylesterase (CE) is implicated in drug metabolism in liver and hepatic steatosis. Deficiency of CE in humans causes obesity, hyperlipidemia, insulin insensitivity, and cancer.

Unit Definition

One unti will hydrolyze one nanomole of 4-nitrophenyl acetate per minute at pH 7.4 at 37°C

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carboxylesterase converts Amplex red to resorufin: Implications for mitochondrial H2O2 release assays
Miwa S, et al.
Free Radical Biology & Medicine, 90, 173-183 (2016)
Non classical monocytes levels, increased by subcutaneous fat-Secretome, are associated with less Rehospitalization after heart failure admission
Eiras S, et al.
Journal of Cardiovascular Translational Research, 10(1), 16-26 (2017)
A carboxylesterase-selective ratiometric fluorescent two-photon probe and its application to hepatocytes and liver tissues
Park SJ, et al.
Chemical Science, 7(6), 3703-3709 (2016)
B Sànchez-Nogué et al.
Environmental science and pollution research international, 20(5), 3480-3488 (2012-12-06)
The common sole, Solea solea (Linneus, 1758), and the Senegalese sole, Solea senegalensis (Kaup, 1858), are two important commercial species that coexist in the NW Mediterranean. In order to assess the species' ability to respond to chemical insults, a comparison
Marie C Fortin et al.
Drug metabolism and disposition: the biological fate of chemicals, 41(2), 326-331 (2012-12-12)
Studies on therapeutic drug disposition in humans have shown significant alterations as the result of pregnancy. However, it is not known whether pesticide metabolic capacity changes throughout pregnancy, which could affect exposure of the developing brain. We sought to determine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service