Skip to Content
Merck
All Photos(1)

Documents

D8440

Sigma-Aldrich

15-Deoxy-Δ12,14-prostaglandin J2

≥95% (HPLC), 1 mg/mL in methyl acetate

Synonym(s):

11-Oxoprosta-(5Z,9,12E,14E)-tetraen-1-oic acid, 15-Deoxy-Δ12,14-PGJ2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28O3
CAS Number:
Molecular Weight:
316.43
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

liquid

concentration

1 mg/mL in methyl acetate

shipped in

wet ice

storage temp.

−20°C

SMILES string

CCCCC\C=C\C=C1/[C@@H](C\C=C/CCCC(O)=O)C=CC1=O

InChI

1S/C20H28O3/c1-2-3-4-5-6-10-13-18-17(15-16-19(18)21)12-9-7-8-11-14-20(22)23/h6-7,9-10,13,15-17H,2-5,8,11-12,14H2,1H3,(H,22,23)/b9-7-,10-6+,18-13+/t17-/m0/s1

InChI key

VHRUMKCAEVRUBK-GODQJPCRSA-N

General description

15-Deoxy-Δ12,14-prostaglandin J2 (15d-PGJ2) is a prostaglandin J2 (PGJ2) metabolite and a naturally occurring derivative of prostaglandin D2. It is produced during inflammatory processes.

Application

15-Deoxy-Δ12,14-prostaglandin J2 has been used:
  • to study its effect on lipid accumulation, viability/mitochondrial activity, and amount of vasculature in vascularized adipose tissue model
  • as a peroxisome proliferator-activated receptor (PPARγ) agonist to activate intestinal fatty acid binding protein (I-FABP)-PPARγ pathway
  • as a supplement in culture medium for induced neural stem/progenitor cells (NSPCs) differentiation

Biochem/physiol Actions

15-Deoxy-Δ12,14-prostaglandin J2 (15d-PGJ2) regulates inflammatory response in vivo. 15d-PGJ2 also elicits PPARγ-independent inhibition of nuclear factor (NF)-κB dependent transcription. It acts as an anti-angiogenic factor and triggers endothelial cell apoptosis.
Selective agonist to PPARγ (peroxisome proliferator-activated receptors). Inhibits the proliferation of cancer cell lines that express PPARγ and cyclooxygenase-2 (COX-2).

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

15.8 °F - closed cup

Flash Point(C)

-9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gas chromatographic-mass spectrometric measurement of 15-deoxy-delta(12,14)-prostaglandin J(2), the peroxisome proliferator-activated receptor gamma ligand, in urine.
C Thévenon et al.
Clinical chemistry, 47(4), 768-770 (2001-03-29)
15-Deoxy-$\Delta$ (12, 14)-prostaglandin J2 induces vascular endothelial cell apoptosis through the sequential activation of MAPKS and p53
Ho TC, et al.
Test, 283(44), 30273-30288 (2008)
Divya Bafna et al.
International journal of molecular sciences, 21(12) (2020-06-18)
Breast cancer (BCa) is one of the most predominantly diagnosed cancers in women. Notably, 70% of BCa diagnoses are Estrogen Receptor α positive (ERα+) making it a critical therapeutic target. With that, the two subtypes of ER, ERα and ERβ
15-deoxy-Delta 12, 14-prostaglandin J2 a prostaglandin D2 metabolite generated during inflammatory processes
Shibata T, et al.
The Journal of biological chemistry, 277(12), 10459-10466 (2002)
N-3 PUFAs attenuate ischemia/reperfusion induced intestinal barrier injury by activating I-FABP-PPAR$\gamma$ pathway
Wang X, et al.
Clinical Nutrition (Edinburgh, Scotland), 31(6), 951-957 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service