Skip to Content
Merck
All Photos(3)

Key Documents

D6679

Sigma-Aldrich

(2,4-Dichlorophenoxy)acetic acid sodium salt monohydrate

suitable for plant cell culture, BioReagent

Synonym(s):

Sodium 2,4-dichlorophenoxyacetate monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H5Cl2NaO3 · H2O
CAS Number:
Molecular Weight:
261.03
MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:
NACRES:
NA.72

product line

BioReagent

Quality Level

form

powder

technique(s)

cell culture | plant: suitable

solubility

H2O: soluble

application(s)

agriculture

SMILES string

O.[Na+].[O-]C(=O)COc1ccc(Cl)cc1Cl

InChI

1S/C8H6Cl2O3.Na.H2O/c9-5-1-2-7(6(10)3-5)13-4-8(11)12;;/h1-3H,4H2,(H,11,12);;1H2/q;+1;/p-1

InChI key

LNSAEWXPCBLGDX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

2,4-Dichlorophenoxyacetic acid (2,4-D) is a synthetic auxin (plant hormone) used in plant cell culture media such as Murashige and Skoog media. 2,4-D is also a widely used herbicide.

Preparation Note

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Control of root hair development in Arabidopsis thaliana by an endoplasmic reticulum anchored member of the R2R3-MYB transcription factor family.
Slabaugh E, Held M, Brandizzi F.
The Plant Journal (2011)
Tomoko Hirano et al.
Plant signaling & behavior, 6(4), 583-585 (2011-03-18)
Fab1/PIKfyve produces Phosphatidylinositol 3,5-bisphosphate (PtdIns (3,5) P2) from Phosphatidylinositol 3-phosphate (PtdIns 3-P), and is involved not only in vacuole/lysosome homeostasis, but also in transporting various proteins to the vacuole or recycling proteins on the plasma membrane (PM) through the use
Alejandra Lara-Chavez et al.
Tree physiology, 31(5), 539-554 (2011-06-04)
The focus of the current project was to establish somatic embryogenesis protocols for the tropical pine species Pinus oocarpa using immature zygotic embryos (ZEs) as explants. Somatic embryogenesis is best supported by mimicking natural seed-embryo developmental conditions, through a tissue
Sibu Simon et al.
Plant science : an international journal of experimental plant biology, 180(3), 454-460 (2011-03-23)
The versatile functionality and physiological importance of the phytohormone auxin is a major focus of attention in contemporary plant science. Recent studies have substantially contributed to our understanding of the molecular mechanisms underlying the physiological role of auxin in plant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service