Skip to Content
Merck
All Photos(1)

Key Documents

C1163

Sigma-Aldrich

Complement C5 deficient serum human

for complement assays

Synonym(s):

Complement serum

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.61

biological source

human

Quality Level

form

solution

technique(s)

activity assay: suitable

UniProt accession no.

storage temp.

−70°C

Gene Information

human ... C5(727)

Application

Complement C5 deficiencies in humans are rare, but often associated with recurrent infections by Neisseria spp. (such as meningitis). Deficiency in C5 results in an impairment of hemolytic activity from both the classical and alternative pathway of the complement system. Recently, genetic research has revealed that a 153 bp deletion in exon 30 results in complete C5 deficiency. Research has also implicated C5 deficiency in a predisposition to cardiac dysfunction when cardiac injury occurs.

Biochem/physiol Actions

Complement C5 deficiency can, paradoxically, protect against certain effects of infection. Patients deficient in C5 are unusually susceptible to meningococcal infection, but have a milder course of infection. Reduced expression of tumor necrosis factor (TNF) may account for this effect.

Physical form

Supplied as a solution in PBS, pH 7.4

Analysis Note

C5 is depleted by immunoadsorption as determined by hemolytic assay.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel role for the fifth component of complement (C5) in cardiac physiology.
Mullick A., et al.
PLoS ONE, 6 (2011)
Magda R Hamczyk et al.
Methods in molecular biology (Clifton, N.J.), 1339, 235-246 (2015-10-09)
The key roles of macrophages in atherosclerosis include the phagocytosis of apoptotic and necrotic cells and cell debris, whose accumulation in atherosclerotic lesions exacerbates inflammation and promotes plaque vulnerability. Evidence is accumulating that macrophage phagocytic functions peak at the early
P A Barton et al.
Infection and immunity, 61(4), 1474-1481 (1993-04-01)
Patients with disseminated Neisseria meningitidis infections (meningococcemia) suffer from a fulminant shock syndrome that is accompanied by extraordinarily high concentrations in serum of tumor necrosis factor (TNF). People with homozygous deficiencies of late complement components (C5, C6, C7, and C8)
P Aguilar-Ramirez et al.
Molecular immunology, 46(10), 2116-2123 (2009-04-21)
The deficiency of complement C5 is rare and frequently associated with severe and recurrent infections, especially caused by Neisseria spp. We observed the absence of component C5 in the serum of 3 siblings from a Brazilian family with history of
Taku Fukuzawa et al.
Scientific reports, 7(1), 1080-1080 (2017-04-26)
Dysregulation of the complement system is linked to the pathogenesis of a variety of hematological disorders. Eculizumab, an anti-complement C5 monoclonal antibody, is the current standard of care for paroxysmal nocturnal hemoglobinuria (PNH) and atypical hemolytic uremic syndrome (aHUS). However

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service