The molecular weight of this product is not determined. Historical data indicate that this material is comprised primarily of the kappa form. In general, Carrageenans have a number average molecular weight range of 193 kDa to 324 kDa with an average molecular weights range of 453 kDa to 652 kDa. Please see the link below for additional information:
https://pubmed.ncbi.nlm.nih.gov/11552743/
C1013
Carrageenan
suitable for gel preparation
Synonym(s):
Vegetable gelatin
About This Item
Recommended Products
biological source
algae (red)
Quality Level
description
Predominantly κ and lesser amounts of λ carrageenan
form
powder
color
white to light brown
useful pH range
7.5-10.5 (1.5% in water)
cation traces
Ca: <3.5%
K: <11%
Na: <2.0%
suitability
suitable for gel preparation
storage temp.
room temp
Looking for similar products? Visit Product Comparison Guide
General description
Application
Preparation Note
Other Notes
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
What is the average molecular weight?
1 answer-
Helpful?
-
-
Is this carrageenan a mixture of kapa and lamda?
1 answer-
Per the properties section, Product C1013 is a mixture of predominantly κ (kappa) and lesser amounts of λ (lambda) carrageenan.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service