Skip to Content
Merck
All Photos(1)

Key Documents

B4558

Sigma-Aldrich

BW B70C

>98% (HPLC), solid

Synonym(s):

N-[3-[3-4(-Fluorophenoxy)phenyl]-1-methyl-2-propenyl]-N-hydroxyurea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17FN2O3
CAS Number:
Molecular Weight:
316.33
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Assay

>98% (HPLC)

form

solid

color

white

solubility

DMSO: soluble 22 mg/mL

storage temp.

2-8°C

SMILES string

CC(\C=C\c1cccc(Oc2ccc(F)cc2)c1)N(O)C(N)=O

InChI

1S/C17H17FN2O3/c1-12(20(22)17(19)21)5-6-13-3-2-4-16(11-13)23-15-9-7-14(18)8-10-15/h2-12,22H,1H3,(H2,19,21)/b6-5+

InChI key

UAIYNMRLUHHRMF-AATRIKPKSA-N

Gene Information

human ... ALOX5AP(241)

Application

BW B70C has been used as a 5-lipoxygenase inhibitor to study its effects on eryptosis in erythrocytes.

Biochem/physiol Actions

BW B70C exhibits anti-inflammatory and anti-tumorigenic activities. It exhibits therapeutic effects against hepatocellular carcinoma. BW B70C plays a role in allergen-induced bronchoconstriction by reducing the allergic airway closure and glycoconjugate secretion.
Selective 5-lipoxygenase inhibitor.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Yeadon et al.
Pulmonary pharmacology, 8(1), 53-63 (1995-02-01)
A method has been established for measurement of tracheal secretions in anaesthetized, ventilated guinea-pigs. The upper trachea was cannulated and perfused with saline. The perfusate was analysed for protein using the Lowry assay and for glycoconjugates ('mucus') by a procedure
Santiago Nahuel Villegas et al.
Cell reports, 22(10), 2541-2549 (2018-03-08)
The PI3K/Akt signaling pathway, Notch, and other oncogenes cooperate in the induction of aggressive cancers. Elucidating how the PI3K/Akt pathway facilitates tumorigenesis by other oncogenes may offer opportunities to develop drugs with fewer side effects than those currently available. Here
Liang Zhang et al.
Environmental toxicology and pharmacology, 56, 145-150 (2017-09-22)
Evidence indicates that Cadmium (Cd) can accumulate in liver, which results in acute or chronic cell damage with unclear complex mechanisms. Thus, we aimed to explore the possible molecules and pathways by using bioinformatics methods Consequently, two datasets (GSE8865 and
B Dugas
Allergie et immunologie, 32(7), 284-289 (2000-11-30)
The effect of Lyprinol was evaluated on LTB4-induced human monocytes (normal and allergic donors) activation. Peripheral blood normal monocyte-derived monocytes when stimulated by Interleukin-4 (IL-4) produced high amounts of leukotriene B4 (LTB4) through the activation of the 5-lipoxygenase pathway. Maximal
T J Morley et al.
Toxicology and applied pharmacology, 146(2), 299-308 (1997-11-05)
In a study of structure-activity relationship with drug-induced nephropathy two lipoxygenase inhibitors, the N-hydroxyurea derivative 70C ((E)-N-{3-[3-(4-fluorophenoxy) phenyl]-1-(R, S)-methylprop-2-enyl}-N-hydroxyurea) and the N-hydroxamic acid analogue 360C ((E)-N-{3-[3-(4-fluorophenoxy) phenyl]-1-(R, S)-methylprop-2-enyl}-N-hydroxamic acid), were administered to rats. 70C and 360C were dosed to female

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service