Skip to Content
Merck
All Photos(3)

Documents

B11001

Sigma-Aldrich

EnPresso® B

Growth system for expressing protein in bacteria

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

sterility

sterile; γ-irradiated

form

tablet

manufacturer/tradename

(BioSilta Oy)

pH

6.8

shipped in

ambient

storage temp.

room temp

General description

Enpresso B by BioSilta Oy

Biochem/physiol Actions

EnPresso® B is a pre-sterilized growth system designed to increase the yield of functional protein from E. coli-based expression systems.

EnPresso® growth systems provide optimal conditions for growth, metabolism and protein expression in microbial cultures. Protein yields are increased by enabling cultures to reach far higher cell densities than those achieved using conventional media. By controlling growth rate and metabolism, a greater proportion of expressed protein can be correctly folded to improve solubility, minimize the risk of inclusion body formation, and ensure functionality of the final product.

EnPresso® growth systems maintain pH, provide adequate minerals, vitamins and trace elements to support growth, and use proprietary EnBase technology to ensure a constant, slow release of glucose from a polysaccharide substrate.

See all available products from EnPresso B Growth Systems.

Physical form

EnPresso® B is supplied in a kit providing sufficient reagents for 20 separate 50 ml cultures. Included in the kit:
40 tablets in 20 white bags
20 tablets in 20 black bags
1 bottle (5 ml) Reagent A

Legal Information

EnBase is a trademark of BioSilta Oy
EnPresso is a registered trademark of BioSilta Oy

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Skin Sens. 1 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jian Li et al.
Journal of biotechnology, 193, 16-22 (2014-12-03)
Heterologous expression of secondary metabolite biosynthesis pathways in a surrogate host, e.g. Escherichia coli, has emerged in recent years as an effective way to produce complex natural products. The nonribosomal peptide (NRP) antibiotic valinomycin has been recombinantly produced in E.
Thomas Horn et al.
Biochimica et biophysica acta, 1831(12), 1702-1713 (2013-08-21)
Mammalian lipoxygenases belong to a family of lipid-peroxidizing enzymes, which have been implicated in cardiovascular, hyperproliferative and neurodegenerative diseases. Here we report that a naturally occurring mutation in the hALOX15 gene leads to expression of a catalytically near-null enzyme variant
Jennifer Jaitzig et al.
ACS synthetic biology, 3(7), 432-438 (2013-12-20)
The structural complexity of nonribosomal peptides (NRPs) impeding economic chemical synthesis and poor cultivability of source organisms limits the development of bioprocesses for novel bioactive compounds. Since nonribosomal peptide synthetases (NRPSs) assemble NRPs from simple amino acid building blocks, heterologous
F Mahboudi et al.
Journal of applied microbiology, 114(2), 364-372 (2012-11-07)
A novel chimeric-truncated form of tissue-type plasminogen activator (t-PA) with improved fibrin affinity and resistance to PAI was successfully produced in CHO expression system during our previous studies. Considering advantages of prokaryotic expression systems, the aim in this study was
Valeria Cafardi et al.
PloS one, 8(11), e81306-e81306 (2013-12-05)
Clostridium difficile is a major cause of infectious diarrhea worldwide. Although the cell surface proteins are recognized to be important in clostridial pathogenesis, biological functions of only a few are known. Also, apart from the toxins, proteins exported by C.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service