Skip to Content
Merck
All Photos(3)

Key Documents

A4529

Sigma-Aldrich

Aprotinin

3-7 TIU/mg solid, lyophilized powder

Synonym(s):

BPTI, Bovine pancreatic trypsin inhibitor, Trasylol, Trypsin inhibitor (basic)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C284H432N84O79S7
CAS Number:
Molecular Weight:
6511.44
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.54
Pricing and availability is not currently available.

Product Name

Aprotinin from bovine lung, lyophilized powder, 3-7 TIU/mg solid

biological source

bovine lung

Quality Level

form

lyophilized powder

specific activity

3-7 TIU/mg solid

mol wt

~6,500

solubility

H2O: >10 mg/mL

UniProt accession no.

storage temp.

2-8°C

SMILES string

S1SCC2NC(=O)CNC(=O)CNC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C5NC(=O)C(NC(=O)CNC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C9N(CCC9)C(=O)CNC(=O)C(NC(=O)C(NC(=O)C%11N(CCC%11)C(=O

InChI key

ZPNFWUPYTFPOJU-UHFFFAOYSA-N

Gene Information

cow ... PTI(404172)

Looking for similar products? Visit Product Comparison Guide

Application

Aprotinin is largely used as an inhibitor of trypsin.

Biochem/physiol Actions

Aprotinin is a competitive serine protease inhibitor that forms stable complexes with and blocks the active sites of enzyme. This binding is reversible, and most aprotinin-protease complexes will dissociate at extreme pH levels >10 or <3. Structurally, Aprotinin is a monomeric globular protein derived from bovine lung that consists of 58 amino acids, arranged in a single polypeptide chain with three crosslinking disulfide bridges.

Unit Definition

One Trypsin Inhibitor Unit (TIU) will decrease the activity of two trypsin units by 50%, where one trypsin unit will hydrolyze 1.0 μmole of N-alpha-benzoyl-DL-arginine p-nitroanilide per minute at pH 7.8 and 25°C. Another commonly used unit is the KIU, with 1 TIU = 1,300 KIU.

Preparation Note

This product is an affinity purified lyophilized powder purified to remove trace impurities. It has an activity of 3-7 TIU/mg.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A M Mahdy et al.
British journal of anaesthesia, 93(6), 842-858 (2004-07-28)
Skilful surgery combined with blood-saving methods and careful management of blood coagulation will all help reduce unnecessary blood loss and transfusion requirements. Excessive surgical bleeding causes hypovolaemia, haemodynamic instability, anaemia and reduced oxygen delivery to tissues, with a subsequent increase
Hemostatic drugs.
P M Mannucci
The New England journal of medicine, 339(4), 245-253 (1998-07-23)
Birte Treeck et al.
The Journal of physiology, 541(Pt 3), 1049-1057 (2002-06-18)
Different changes in glomerular filtration rates (GFR) in deep and superficial glomeruli have been suggested to influence renal NaCl excretion and concentrating ability. Angiotensin II (AngII) has been implicated in such changes, but the experimental evidence has been conflicting, probably
D Baran et al.
Acta physiologica (Oxford, England), 186(3), 209-221 (2006-02-25)
The renal tubular uptake of 125I-Aprotinin (*Ap) is on average located more superficially than its filtration site, causing transfer of some of *Ap filtered in deep to more superficial cortical zones. 125I-Cystatin C (*Cy) showed less uptake in deep cortical
Ilaria Musante et al.
Experimental physiology, 104(6), 866-875 (2019-03-30)
What is the central question of this study? What is the precise subcellular localization of the epithelial sodium channel (ENaC) in human airway epithelium? What is the main finding and its importance? ENaC protein has an unexpected localization in the

Articles

While aprotinin and bovine pancreatic trypsin inhibitor (BPTI) are the same protein sequence, the term aprotinin is typically used when describing the protein derived from bovine lung.

Protocols

Objective: To standardize a procedure for the enzymatic assay of Aprotinin.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service