Skip to Content
Merck
All Photos(4)

Documents

A2929

Sigma-Aldrich

Agarose

For pulsed field electrophoresis running gel

Synonym(s):

3,6-Anhydro-α-L-galacto-β-D-galactan

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41105317
PubChem Substance ID:
NACRES:
NA.25

biological source

algae (red)

form

powder

technique(s)

electrophoresis: suitable

impurities

≤10% water

EEO

≤0.10

transition temp

gel point 38-43 °C (10% gel)

gel strength

≥2000 g/cm2

anion traces

sulfate (SO42-): ≤0.20%

foreign activity

DNase, RNase, NICKase, none detected

InChI

1S/C24H38O19/c25-1-5-9(27)11(29)12(30)22(38-5)41-17-8-4-36-20(17)15(33)24(40-8)43-18-10(28)6(2-26)39-23(14(18)32)42-16-7-3-35-19(16)13(31)21(34)37-7/h5-34H,1-4H2/t5-,6-,7+,8+,9+,10+,11+,12-,13+,14-,15+,16-,17-,18+,19+,20+,21-,22+,23+,24+/m1/s1

InChI key

MJQHZNBUODTQTK-WKGBVCLCSA-N

Looking for similar products? Visit Product Comparison Guide

Specificity

Suitable for the separation of high molecular weight DNA. Gels are easy to handle and give faster separation and better resolution of high molecular weight DNA by field inversion electrophoresis than our routine-use agarose.

Application

Especially formulated for fast resolution of large DNA (10-2000 ·kb) by pulsed-field gel electrophoresis (PFGE) or conventional electrophoresis. Pulsed-field gel electrophoresis was developed in 1984 to deal with "reptation" of large DNA′s whose size surpass the limit of resolution and all migrate at the same rate. Alternately pulsed, orthogonally oriented electric fields are applied across the gel. Every time the direction of the electric field changes, the large DNA′s can no longer migrate until they become reoriented along the new electric field. The time required for reorientation is dependent on the size of the DNA, therefore the DNA is fractionated by size.

Features and Benefits

  • Low EEO allows for increased voltages and more rapid DNA migration
  • Greater gel strength allows for stability of lower concentration gels leading to faster DNA migration
  • Gels exhibit low background fluorescence using ethidium bromide staining

Analysis Note

The following is a list of properties associated with our agaroses:
Sulfate content - used as an indicator of purity, since sulfate is the major ionic group present.
Gel strength - the force that must be applied to a gel to cause it to fracture.
Gel point - the temperature at which an aqueous agarose solution forms a gel as it cools. Agarose solutions exhibit hysteresis in the liquid-to-gel transition - that is, their gel point is not the same as their melting temperature.
Electroendosmosis (EEO) - a movement of liquid through the gel. Anionic groups in an agarose gel are affixed to the matrix and cannot move, but dissociable counter cations can migrate toward the cathode in the matrix, giving rise to EEO. Since electrophoretic movement of biopolymers is usually toward the anode, EEO can disrupt separations because of internal convection.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sifang Liao et al.
Frontiers in endocrinology, 11, 180-180 (2020-05-07)
The insulin/IGF-signaling pathway is central in control of nutrient-dependent growth during development, and in adult physiology and longevity. Eight insulin-like peptides (DILP1-8) have been identified in Drosophila, and several of these are known to regulate growth, metabolism, reproduction, stress responses
Yu-Hsuan Cheng et al.
BMC biology, 18(1), 180-180 (2020-12-01)
Ciliates are an ancient and diverse eukaryotic group found in various environments. A unique feature of ciliates is their nuclear dimorphism, by which two types of nuclei, the diploid germline micronucleus (MIC) and polyploidy somatic macronucleus (MAC), are present in
Gianluca Vadalà et al.
Spine, 38(6), E319-E324 (2013-01-18)
Descriptive anatomical study on ovine and human cadaveric lumbar spinal segments. To describe the alternative transpedicular approach to deliver therapeutic agents into intervertebral disc (IVD). The present delivery approach of therapeutic agents (growth factors/cells/hydrogels) within the IVD is through injection
Víctor Carriel et al.
Journal of neural engineering, 10(2), 026022-026022 (2013-03-27)
The objective was to study the effectiveness of a commercially available collagen conduit filled with fibrin-agarose hydrogels alone or with fibrin-agarose hydrogels containing autologous adipose-derived mesenchymal stem cells (ADMSCs) in a rat sciatic nerve injury model. A 10 mm gap
Maria Carmen Piñon et al.
The Journal of physiology, 587(Pt 9), 1903-1915 (2009-03-18)
In the Golli-tau-eGFP (GTE) transgenic mouse the reporter gene expression is largely confined to the layer of subplate neurons (SPn), providing an opportunity to study their intracortical and extracortical projections. In this study, we examined the thalamic afferents and layer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service