Skip to Content
Merck
All Photos(4)

Documents

A2169

Sigma-Aldrich

3′-Azido-3′-deoxythymidine

≥98% (HPLC), powder, reverse transcriptase inhibitor

Synonym(s):

AZT, Azidothymidine, ZDV, Zidovudine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N5O4
CAS Number:
Molecular Weight:
267.24
Beilstein:
3595791
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

3′-Azido-3′-deoxythymidine, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

mp

113-115 °C (lit.)

solubility

H2O: 50 mg/mL

storage temp.

−20°C

SMILES string

CC1=CN([C@H]2C[C@H](N=[N+]=[N-])[C@@H](CO)O2)C(=O)NC1=O

InChI

1S/C10H13N5O4/c1-5-3-15(10(18)12-9(5)17)8-2-6(13-14-11)7(4-16)19-8/h3,6-8,16H,2,4H2,1H3,(H,12,17,18)/t6-,7+,8+/m0/s1

InChI key

HBOMLICNUCNMMY-XLPZGREQSA-N

Gene Information

human ... HIVE1(3095)
mouse ... Slc29a1(63959)

Looking for similar products? Visit Product Comparison Guide

Application

3′-Azido-3′-deoxythymidine has been used as the reverse transcriptase inhibitor to inhibit the viral genome integration and as an antimicrobial agent to evaluate its anti-Salmonella activity.

Biochem/physiol Actions

Azidothymidine (AZT), a thymidine analogue, is a reverse transcriptase inhibitor against the HIV-1 virus. Azidothymidine has been shown to decrease CRISPR-mediated homology-directed repair (HDR) efficiency.
Reverse transcriptase inhibitor active against HIV-1 virus.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Muta. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

In vitro activities of nucleoside analog antiviral agents against salmonellae.
Sperber S J, et al.
Antimicrobial Agents and Chemotherapy, 37(1), 106-110 (1993)
Protein overexpression following lentiviral infection of primary mature neutrophils is due to pseudotransduction
Geering B, et al.
Journal of Immunological Methods, 373(1-2), 209-218 (2011)
Critically short telomeres derepress retrotransposons to promote genome instability in embryonic stem cells.
Zhao, et al.
Cell discovery, 9, 45-45 (2023)
Markus Moll et al.
European journal of immunology, 39(3), 902-911 (2009-02-07)
Invariant CD1d-restricted NKT cells play important roles in regulating both innate and adaptive immunity. They are targeted by HIV-1 infection and severely reduced in number or even lost in many infected subjects. Here, we have investigated the characteristics of NKT
Aoife G Cotter et al.
The Journal of clinical endocrinology and metabolism, 98(4), 1659-1666 (2013-02-26)
In virologically suppressed, antiretroviral-treated patients, the effect of switching to tenofovir (TDF) on bone biomarkers compared to patients remaining on stable antiretroviral therapy is unknown. We examined bone biomarkers (osteocalcin [OC], procollagen type 1 amino-terminal propeptide, and C-terminal cross-linking telopeptide

Articles

Modulation of homology-directed repair (HDR) within the context of CRISPR-genome editing has led to the identification of small molecules that enhance CRISPR-mediated HDR efficiency in various cell types.

Modulation of homology-directed repair (HDR) within the context of CRISPR-genome editing has led to the identification of small molecules that enhance CRISPR-mediated HDR efficiency in various cell types.

Modulation of homology-directed repair (HDR) within the context of CRISPR-genome editing has led to the identification of small molecules that enhance CRISPR-mediated HDR efficiency in various cell types.

Modulation of homology-directed repair (HDR) within the context of CRISPR-genome editing has led to the identification of small molecules that enhance CRISPR-mediated HDR efficiency in various cell types.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service