Skip to Content
Merck
All Photos(3)

Key Documents

A1882

Sigma-Aldrich

6-Azauridine

Synonym(s):

2-β-D-Ribofuranosyl-1,2,4-triazine-3,5(2H,4H)-dione, 6-Azauracil riboside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11N3O6
CAS Number:
Molecular Weight:
245.19
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Assay

≥99% (TLC)

form

powder

mp

157-159 °C (lit.)

solubility

water: 50 mg/mL, clear, colorless

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N2N=CC(=O)NC2=O

InChI

1S/C8H11N3O6/c12-2-3-5(14)6(15)7(17-3)11-8(16)10-4(13)1-9-11/h1,3,5-7,12,14-15H,2H2,(H,10,13,16)/t3-,5-,6-,7-/m1/s1

InChI key

WYXSYVWAUAUWLD-SHUUEZRQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

6-Azauridine is a pyrimidine nucleoside analog.

Application

6-Azauridine (AzUrd) blocks the conversion of orotic acid into UMP and it is used in antiviral studies.
6-Azauridine has been used:
  • in uridine monophosphate synthase (UMPS) activity assay
  • as an antiviral agent to study its inhibition effect and cytotoxic potential on foot and mouth disease virus
  • to screen for anticryptosporidial activity
  • in the pretreatment of HeLa cells to study its effect on inhibition of cellular uridine synthesis before and during chase with 5-bromouridine 5′-triphosphate (BrUTP)
  • as a reference compound for comparing the antiviral activity and cytotoxic activity against viral host cell lines

Biochem/physiol Actions

6-azauridine is a prodrug, upon conversion to 6-aza-UMP, inhibits uridine monophosphate synthase (UMPS). It is a broad spectrum anti-metabolite. It interferes with pyrimidine biosynthesis and affects the cellular nucleic acid levels. It is considered as a antineoplastic metabolite. It inhibits RNA viruses like chikungunya virus, semliki forest virus, and human coronavirus. It significantly inhibits the growth of Cryptosporidium parasites.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Bhasin et al.
The American journal of physiology, 243(3), E234-E239 (1982-09-01)
Ketonemic states complicating late pregnancy are accompanied by lower brain weights in the newborn. Potential mechanisms whereby ketone bodies might inhibit cell proliferation were therefore examined in the fetal rat brain slice by measuring their impact on the de novo
Jean Marc Crance et al.
Antiviral research, 58(1), 73-79 (2003-04-30)
Ribavirin, interferon-alpha (IFN-alpha), 6-azauridine and glycyrrhizin were tested in vitro for their antiviral activities against 11 pathogenic flaviviruses belonging to principal antigenic complexes or individual serogroups of medical importance: dengue, Japanese encephalitis, mammalian tick-borne and yellow fever virus (YFV) groups.
Takashi Kobayashi et al.
Physical chemistry chemical physics : PCCP, 12(19), 5140-5148 (2010-05-07)
Excited state characteristics of 6-azauridine (6AUd), which is known as a medicine against psoriasis and neoplastic, were investigated with laser plash photolysis, time-resolved thermal lensing, and near IR single photon counting method. The triplet-triplet absorption spectrum of 6AUd was observed
Donald F Smee et al.
Journal of virological methods, 246, 51-57 (2017-04-01)
Studies were conducted to determine the performance of four dyes in assessing antiviral activities of compounds against three RNA viruses with differing cytopathogenic properties. Dyes included alamarBlue
Krzysztof Pyrc et al.
Antimicrobial agents and chemotherapy, 50(6), 2000-2008 (2006-05-26)
Human coronavirus NL63 (HCoV-NL63), a recently discovered member of the Coronaviridae family, has spread worldwide and is associated with acute respiratory illness in young children and elderly and immunocompromised persons. Further analysis of HCoV-NL63 pathogenicity seems warranted, in particular because

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service