Skip to Content
Merck
All Photos(1)

Key Documents

84959

Sigma-Aldrich

L-Serine

≥99.5% (NT), suitable for cell culture, BioUltra

Synonym(s):

(S)-2-Amino-3-hydroxypropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
105.09
Beilstein:
1721404
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26
Pricing and availability is not currently available.

Product Name

L-Serine, BioUltra, ≥99.5% (NT)

product line

BioUltra

Quality Level

Assay

≥99.5% (NT)

form

powder or crystals

optical activity

[α]20/D +13.5±0.5°, c = 5% in 5 M HCl

technique(s)

cell culture | mammalian: suitable

impurities

insoluble matter, passes filter test
≤0.3% foreign amino acids

ign. residue

≤0.1% (as SO4)

loss

≤0.1% loss on drying, 20 °C (HV)

color

white

mp

222 °C (dec.) (lit.)

solubility

1 M HCl: 0.5 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
NH4+: ≤100 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

0.5 M in 1 M HCl

UV absorption

λ: 260 nm Amax: 0.05
λ: 280 nm Amax: 0.05

SMILES string

N[C@@H](CO)C(O)=O

InChI

1S/C3H7NO3/c4-2(1-5)3(6)7/h2,5H,1,4H2,(H,6,7)/t2-/m0/s1

InChI key

MTCFGRXMJLQNBG-REOHCLBHSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Effect of SlSAHH2 on metabolites in over-expressed and wild-type tomato fruit.: Research focused on the metabolite variations in genetically modified tomatoes, including the impact of L-Serine in metabolic engineering to enhance nutritional values and stress resistance (Yang et al., 2024).

Biochem/physiol Actions

Precursor of glycine by serine hydroxymethyltransferase.

Other Notes

Synthesis of labels for protonation studies by Fourier Transform infrared methods[1]; Stabilizes mung bean nuclease at pH values below seven[2]; Amino acid spacing in isotachophoresis on polyacrylamide gel - a critical evaluation[3]; Growth requirement of various microorganisms[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

CRC Handbook of Microbiology, 4, 1-1 (1974)
Bacteriorhodopsin: fourier transform infrared methods for studies of protonation of carboxyl groups.
G Dollinger et al.
Methods in enzymology, 127, 649-662 (1986-01-01)
Amino acid spacing in isotachophoresi; on polyacrylamide gel: a critical evaluation.
N Y Nguyen et al.
Analytical biochemistry, 94(1), 202-210 (1979-04-01)
Theory of protein solubility.
T Arakawa et al.
Methods in enzymology, 114, 49-77 (1985-01-01)
Miyoshi Haruta et al.
Science (New York, N.Y.), 343(6169), 408-411 (2014-01-25)
Plant cells are immobile; thus, plant growth and development depend on cell expansion rather than cell migration. The molecular mechanism by which the plasma membrane initiates changes in the cell expansion rate remains elusive. We found that a secreted peptide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service