The composition of this aqueous solution is considered proprietary.
56796
Micro particles based on silicon dioxide
size: 0.5 μm
Synonym(s):
Beads based on silicon dioxide, microsize, Silicon dioxide beads
About This Item
Recommended Products
grade
analytical standard
Quality Level
form
aqueous suspension
concentration
5% (solids)
particle size
0.5 μm
Mw/Mn
0.15
application(s)
glass & ceramic
industrial qc
pharmaceutical
format
neat
storage temp.
2-8°C
SMILES string
O=[Si]=O
InChI
1S/O2Si/c1-3-2
InChI key
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Also used to fabricate silicon-based nano-electro-mechanical systems (NEMS) for mass sensing application.[1]
Features and Benefits
- suitable for routine instrument calibration checks, testing and corrections
- available in 5 and 10 mL pack sizes as neat samples
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
SiO2 sphere가 담겨있는 aqueous solution 의 구성 요소를 알 수 있을까요?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service