Skip to Content
Merck
All Photos(1)

Key Documents

56250

Sigma-Aldrich

trans-4-Hydroxy-L-proline

≥99.0% (NT), BioXtra

Synonym(s):

(2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid, Hyp

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO3
CAS Number:
Molecular Weight:
131.13
Beilstein:
81441
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

trans-4-Hydroxy-L-proline, BioXtra, ≥99.0% (NT)

product line

BioXtra

Quality Level

Assay

≥99.0% (NT)

form

solid

optical activity

[α]20/D −76.0±1.5°, c = 5% in H2O

impurities

≤0.3% foreign amino acids

ign. residue

≤0.1% (as SO4)

loss

≤0.1% loss on drying, 110 °C

color

colorless to white

mp

273 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
NH4+: ≤100 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

application(s)

peptide synthesis

SMILES string

O[C@H]1CN[C@@H](C1)C(O)=O

InChI

1S/C5H9NO3/c7-3-1-4(5(8)9)6-2-3/h3-4,6-7H,1-2H2,(H,8,9)/t3-,4+/m1/s1

InChI key

PMMYEEVYMWASQN-DMTCNVIQSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Trans-4-Hydroxy-L-proline is used as a starting material in the synthesis of (-)-secrinine; the synthesis of prolinol-based nucleotide analogues with N-phosphonomethyl moiety attached to the nitrogen atom of prolinol ring and in asymmetric synthesis of the ABC-ring system of the antitumor antibiotic MPC1001.
Trans-4-Hydroxy-L-proline is used in the organic synthesis of depsilairdin analogues, noncompetitive peptidomimetic inhibitors of multidrug resistance P-glycoprotein and 3-guaninyl-5-hydroxymethyl-2-pyrrolidinone (4) or 3-adeninyl-5-hydroxymethyl-2-pyrrolidinone (5) nucleoside analogues.

Other Notes

Natural constituent of animal structural proteins such as collagen and elastin.
Enzymes and intermediates of hydroxyproline degradation

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

E. Adams
Methods in Enzymology, 17B, 266-266 (1971)
Hsuan-Shu Lee et al.
Journal of gastroenterology and hepatology, 20(7), 1109-1114 (2005-06-16)
Needle liver biopsy is essential for the diagnosis of liver fibrosis and cirrhosis. However, sampling variability has been reported by semiquantitative methods. The aim of the present study was to investigate quantitatively the sampling variability of needle liver biopsy on
Ville V Meretoja et al.
Biomaterials, 34(17), 4266-4273 (2013-03-16)
In this work, we investigated the effects of lowered oxygen tension (20% and 5% O2) on the chondrogenesis and hypertrophy of articular chondrocytes (ACs), mesenchymal stem cells (MSCs) and their co-cultures with a 30:70 AC:MSC ratio. Cells were cultured for
Shahrzad Hosseininia et al.
BMC musculoskeletal disorders, 14, 18-18 (2013-01-11)
It remains to be shown whether OA shares molecular similarities between different joints in humans. This study provides evidence for similarities in cartilage molecular damage in osteoarthritic (OA) joints. Articular cartilage from osteoarthritic hip joints were analysed and compared to
Jacob Kofoed et al.
Chemical communications (Cambridge, England), (14)(14), 1482-1484 (2006-04-01)
The aldol reaction of acetone with aldehydes in aqueous medium under catalysis by zinc-proline (Zn(L-Pro)2) and secondary amines such as proline, (2S,4R)-4-hydroxyproline (Hyp) and (S)-(+)-1-(2-pyrrolidinomethyl)pyrrolidine (PMP) is shown to proceed by an enamine mechanism, as evidenced by reductive trapping of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service