Skip to Content
Merck
All Photos(1)

Key Documents

49937

Sigma-Aldrich

Atto 488-Streptavidin

BioReagent, suitable for fluorescence

Synonym(s):

Streptavidin-Atto 488

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161900
NACRES:
NA.25

product line

BioReagent

form

solid

manufacturer/tradename

ATTO-TEC GmbH

λ

in PBS, pH 7.4

UV absorption

λ: 500-506 nm Amax

suitability

corresponds for gel electrophoresis
suitable for fluorescence

storage temp.

−20°C

Analysis Note

dye-to-protein ratio ≥1.5

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Breaking the Diffraction Barrier in Fluorescence Microscopy by Optical Shelving.
Bretschneider, S.; Eggeling, Chr.; Hell, S. W.
Physical Review Letters, 98(21) (2007)
Instant Live-Cell Super-Resolution Imaging of Cellular Structures by Nanoinjection of Fluorescent Probes.
Hennig, S.; et al.
Nano Letters, 15(2), 1374-1381 (2015)
Dual color localization microscopy of cellular nanostructures.
Gunkel, M.; et al.
Biotechnology Journal, 4(6), 927-938 (2009)
Dynamic Superresolution Imaging of Endogenous Proteins on Living Cells at Ultra-High Density.
Giannone, G.; et al.
Biophysical Journal, 99(4), 1303-1310 (2010)
The changing point-spread function. Single-molecule-based super-resolution imaging.
Horrocks, M. H.; Palayret, M.; Klenerman, D.; Lee, S. F.
Histochemistry and Cell Biology, 141(6), 577-585 (2014)

Articles

Atto dyes are a series of fluorescent dyes that meet the critical needs of modern fluorescent technologies.

Atto dyes are a series of fluorescent dyes that meet the critical needs of modern fluorescent technologies.

Atto dyes are a series of fluorescent dyes that meet the critical needs of modern fluorescent technologies.

Atto dyes are a series of fluorescent dyes that meet the critical needs of modern fluorescent technologies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service