Skip to Content
Merck
All Photos(1)

Key Documents

44054

Supelco

Micro particles based on silicon dioxide

size: 5 μm

Synonym(s):

Silica dioxide microsphere (5 μm), Micro silicon (5 μm), Beads based on silicon dioxide, microsize, Silicon dioxide beads

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

form

aqueous suspension

concentration

5% (solids)

particle size

5 μm std dev ≤0.35 μm

application(s)

glass & ceramic
industrial qc
pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silicon dioxide based microparticles are ideal to determine particle size distribution (PSD) profile of test samples.
The particle diameter of 5 μm is measured using Coulter Multisizer.

Application

Used for regular calibration and checking of particle size instruments. Also used:
  • to study transport properties of microscopic particles via heterogeneous media[1]
  • as a test standard to validate denoising method used in optical diffraction tomography[2]
  • in accelerated annealing experiments involving colloidal silica monolayers[3]
  • to evaluate the measurement accuracy of color-coded LED microscopy (cLEDscope) technique[4]
  • as a calibration standard in optical trapping system to measure surface mechanical properties[5]

Features and Benefits

  • suitable for routine instrument calibration monitoring, testing and corrections
  • particle size traceable to Community Bureau of Reference (BCR) standards
  • available in 5 mL pack size as a neat sample

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Single-exposure quantitative phase imaging in color-coded LED microscopy
Lee W, et al.
Optics Express, 25(7), 8398-8411 (2017)
Cycle-consistent deep learning approach to coherent noise reduction in optical diffraction tomography
Choi G, et al.
Optics Express, 27(4), 4927-4943 (2019)
Sergi Granados Leyva et al.
Soft matter, 16(30), 6985-6992 (2020-07-17)
We combine experiments and numerical simulations to investigate the emergence of clogging in a system of interacting paramagnetic colloidal particles driven against a disordered landscape of larger obstacles. We consider a single aperture in a landscape of immobile silica particles
Chansuk Park et al.
Journal of the Optical Society of America. A, Optics, image science, and vision, 35(11), 1891-1898 (2018-11-22)
Optical diffraction tomography (ODT) is a three-dimensional (3D) quantitative phase imaging technique, which enables the reconstruction of the 3D refractive index (RI) distribution of a transparent sample. Due to its fast, non-invasive, and quantitative imaging capability, ODT has emerged as
Sophie Ramananarivo et al.
Nature communications, 10(1), 3380-3380 (2019-07-31)
Molecular motors are essential to the living, generating fluctuations that boost transport and assist assembly. Active colloids, that consume energy to move, hold similar potential for man-made materials controlled by forces generated from within. Yet, their use as a powerhouse

Questions

1–3 of 3 Questions  
  1. What kind of methods/protocol is used to fabricate these silica microparticles? and is it bioactive with cells?

    1 answer
    1. The preparation method is considered proprietary. Information on bioactivity with cells is not available.

      Helpful?

  2. Are these Silica particles partly porous? What is the approximate refractive index?

    1 answer
    1. These beads are considered to be non-porous. BET experiments with comparable particles revealed practically no inner surface. As mentioned in the PROPERTIES section, the particle size is 5 μm with a standard deviation of ≤0.35 μm.

      Helpful?

  3. Are these silica particles charged? If yes, the what is surface charge density of these particles.

    1 answer
    1. The silica particles have not been tested for the presence or absence of a charge.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service