Skip to Content
Merck
All Photos(1)

Key Documents

38733

Sigma-Aldrich

Laemmli Lysis-buffer

non smelling

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161700
NACRES:
NA.25

form

liquid

composition

bromophenol blue, 0.004%
DTT, 400 mM
glycerol, 20%
SDS, 4%
TRIS, 0.125 M

pH

~6.8

suitability

in accordance for electrophoresis test

storage temp.

−20°C

Application

Laemmli Lysis-buffer is an non-smelling lysis buffer used to prepare protein samples for SDS gel electrophoresis.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vincent Archambault et al.
Genes & development, 22(19), 2707-2720 (2008-10-04)
The conserved Polo kinase controls multiple events in mitosis and cytokinesis. Although Polo-like kinases are regulated by phosphorylation and proteolysis, control of subcellular localization plays a major role in coordinating their mitotic functions. This is achieved largely by the Polo-Box
Sean R Gallagher
Current protocols in protein science, Chapter 10, Unit 10-Unit 10 (2012-04-04)
One-dimensional gel electrophoresis of proteins provides information about the molecular size, amount, and purity of a protein sample. Separated proteins can be recovered from polyacrylamide gels for subsequent characterization by a variety of secondary techniques, such as mass spectrometry to
Xiaoqiang Wu et al.
Analytical biochemistry, 421(1), 347-349 (2011-11-01)
A modified Laemmli sodium dodecyl sulfate-polyacrylamide gel electrophoresis (SDS-PAGE) protocol is described. The new method saves 30 min for gel casting without loss of the resolution power of Laemmli gel. In this method, both the upper and lower gels can
Sean R Gallagher
Current protocols in immunology, Chapter 8, Unit 8-Unit 8 (2008-04-25)
Electrophoresis is used to separate complex mixtures of proteins (e.g., from cells, subcellular fractions, column fractions, or immunoprecipitates), to investigate subunit compositions, and to verify homogeneity of protein samples. It can also serve to purify proteins for use in further
K Zajda et al.
Human & experimental toxicology, 39(3), 276-289 (2019-11-09)
Tissue-dependent oestrogenic and anti-oestrogenic activity of polycyclic aromatic hydrocarbons (PAHs) has been suggested. In this study, the effect of two PAH mixtures, M1 composed of all 16 priority pollutants and M2 composed of five (noted in the highest levels) compounds

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service