Skip to Content
Merck
All Photos(2)

Key Documents

36679

Supelco

PCB No 31

analytical standard

Synonym(s):

2,4′,5-Trichlorobiphenyl, 2,4′,5-PCB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H7Cl3
CAS Number:
Molecular Weight:
257.54
Beilstein:
1961529
Ballschmiter Number:
31
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

SMILES string

Clc1ccc(cc1)-c2cc(Cl)ccc2Cl

InChI

1S/C12H7Cl3/c13-9-3-1-8(2-4-9)11-7-10(14)5-6-12(11)15/h1-7H

InChI key

VAHKBZSAUKPEOV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Zou et al.
Ecotoxicology and environmental safety, 38(3), 281-285 (1998-02-21)
The effects of five xenobiotics, 2,4,5-trichloribiphenyl (PCB29), the polychlorinated biphenyl (PCB) Aroclor 1242, diethyl phthalate, lindane, and 4-octylphenol, on molting of Daphnia magna were investigated. All except PCB29 are known to have unexpected estrogenicity in vertebrates. Daphnids exposed to PCB29
Sachi Taniyasu et al.
Environmental pollution (Barking, Essex : 1987), 126(2), 169-178 (2003-08-21)
Technical polychlorinated byphenyl (PCB) preparations--Delors 103, 104, 105, and 106--produced in the former Czechoslovakia were analyzed for their chlorobiphenyl (CB), chloronaphthalene (PCN) and chlorinated dibenzofuran (PCDF) composition and content using high resolution gas chromatography-mass spectrometry techniques. The congener patterns of
J Rehulka et al.
Diseases of aquatic organisms, 62(1-2), 147-153 (2005-01-15)
Rainbow trout at a weight of 223+/-12 g (mean+/-SD) were experimentally injected with a technical mixture of Delor 103 to evaluate the red blood cell indices (red blood cell count, haematocrit, haemoglobin, mean corpuscular volume, mean corpuscular haemoglobin, mean corpuscular
I Kalina et al.
Teratogenesis, carcinogenesis, and mutagenesis, 11(2), 77-82 (1991-01-01)
The ability of polychlorinated biphenyls (PCBs) to induce chromosome aberrations and sister chromatid exchanges (SCE) in peripheral lymphocytes was studied in a group of workers occupationally exposed to PCBs during the production of the Czechoslovak PCB products Delor 103 and
Ewa Lucia Gregoraszczuk et al.
Endocrine regulations, 39(2), 33-41 (2005-10-19)
To compare the effects on ovarian function of low chlorinated polychlorinated biphenyls (PCBs) mixture generally distributed at a faster rate and metabolised to a greater extent (Delor 103) with high chlorinated PCBs mixture which are accumulated in large amounts in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service