Skip to Content
Merck
All Photos(4)

Key Documents

901230

Sigma-Aldrich

Ammonium chloride

anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%

Synonym(s):

Salmiac

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4Cl
CAS Number:
Molecular Weight:
53.49
Beilstein:
4371014
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21
Assay:
≥99.5%
grade:
ACS reagent
anhydrous
form:
powder or crystals
Pricing and availability is not currently available.

grade

ACS reagent
anhydrous

Quality Level

Agency

suitable for SM 4500 - NH3
suitable for SM 5210

vapor density

1.9 (vs air)

vapor pressure

1 mmHg ( 160.4 °C)

product line

Redi-Dri

Assay

≥99.5%

form

powder or crystals

impurities

≤0.005% insolubles

ign. residue

≤0.01%

pH

4.5-5.5 (25 °C, 5%)

mp

340 °C (subl.) (lit.)

anion traces

phosphate (PO43-): ≤2 ppm
sulfate (SO42-): ≤0.002%

cation traces

Ca: ≤0.001%
Fe: ≤2 ppm
Mg: ≤5 ppm
heavy metals: ≤5 ppm (by ICP)

SMILES string

N.Cl

InChI

1S/ClH.H3N/h1H;1H3

InChI key

NLXLAEXVIDQMFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Crystals of ammonium chloride (NH4Cl) belong to the cubic crystal system. It can be synthesized by reacting HCl with aqueous ammonia solutions. NH4Cl is formed as a by-product during soda manufactured by Solvay process.[1]

Application

Ammonium chloride (NH4Cl) has been used as one of the components of red blood cell (RBC) lysis buffer for use in various studies.[2][3] It may be used as a catalyst in the preparation 3,4-dihydropyrimidin-2-(1H)-ones.[4]
Inorganic ammonium ions (NH4+) effectively catalyze the aldol condensation of carbonyl compounds.[5] It serves as a source of nitrogen during the preparation of trimethylamine hydrochloride (TMA·HCl).[6]

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Immune Cell Isolation from Mouse Femur Bone Marrow.
Liu X and Quan N.
Bio-protocol, 5(20) (2015)
Mark E Kusek et al.
Journal of visualized experiments : JoVE, (83)(83), e50823-e50823 (2014-01-17)
Mucosal surfaces serve as protective barriers against pathogenic organisms. Innate immune responses are activated upon sensing pathogen leading to the infiltration of tissues with migrating inflammatory cells, primarily neutrophils. This process has the potential to be destructive to tissues if
Selective Synthesis of Trimethylamine by Catalytic N-Methylation of Ammonia and Ammonium Chloride by utilizing Carbon Dioxide and Molecular Hydrogen.
Beydoun K, et al.
ChemCatChem, 8(1), 135-138 (2016)
Barbara Nozière et al.
Physical chemistry chemical physics : PCCP, 12(15), 3864-3872 (2010-04-02)
In natural environments such as atmospheric aerosols, organic compounds coexist with inorganic salts but, until recently, were not thought to interact chemically. We have recently shown that inorganic ammonium ions, NH(4)(+), act as catalysts for acetal formation from glyoxal, a
Ammonium chloride-catalyzed one-pot synthesis of 3,4-dihydropyrimidin-2-(1H)-ones under solvent-free conditions
Shaabani A, et al
Tetrahedron Letters, 44, 857-859 (2003)

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service