Skip to Content
Merck
All Photos(1)

Documents

383139

Sigma-Aldrich

Sodium bismuthate

ACS reagent

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaBiO3
CAS Number:
Molecular Weight:
279.97
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24

grade

ACS reagent

description

oxidizing efficiency >= 99.6%

Assay

≥80.0%

form

powder

reaction suitability

reagent type: catalyst
core: bismuth

anion traces

chloride (Cl-): ≤0.002%

cation traces

Mn: ≤5 ppm

SMILES string

[Na+].[O-][Bi](=O)=O

InChI

1S/Bi.Na.3O/q;+1;;;-1

InChI key

PNYYBUOBTVHFDN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sodium bismuthate may be used in the following processes:
  • As an oxidant in the preparation of hexavalent Americium (Am) during the separation of Am from the lanthanides.
  • Hydrothermal synthesis of bismuth oxychloride (BiOCl) micrometer-sized single crystals.
  • As a precursor in the synthesis of bismuth nanoforms under microwave (MW) irradiation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ligand-Free One-Step Synthesis of {001} Faceted Semiconducting BiOCl Single Crystals and Their Photocatalytic Activity.
Biswas A, et al.
Crystal Growth & Design, 14(1), 236-239 (2013)
Correlation of crystal structures and electronic structures with visible light photocatalytic properties of NaBiO3.
Liu J, et al.
Chemical Physics Letters, 572, 101-105 (2013)
A comparison of bismuth nanoforms obtained in vacuum and air by microwave heating of bismuth powder.
Kharissova OV, et al.
Materials Chemistry and Physics, 121(3), 489-496 (2010)
Characterizing Diamylamylphosphonate (DAAP) as an Americium Ligand for Nuclear Fuel-Cycle Applications.
Mincher BJ, et al.
Solvent Extr. Ion Exch., 32(2), 153-166 (2014)
Kai Yu et al.
Environmental science & technology, 46(13), 7318-7326 (2012-05-24)
Organic dye degradation was achieved via direct oxidation by bismuth silver oxide coupled with visible light photocatalysis by sodium bismuthate. Crystal violet dye decomposition by each reagent proceeded via two distinct pathways, each involving different active oxygen species. A comparison

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service